MABF156A
Anti-CD44, (human/mouse), violetFluor® 450, clone IM7 Antibody
clone IM7, from rat, violetFluor® 450
Synonym(s):
CD44 antigen, Extracellular matrix receptor III (ECMR-III), GP90 lymphocyte homing/adhesion receptor, HUTCH-I, Hermes antigen, Hyaluronate receptor, Lymphocyte antigen 24 (Ly-24), Phagocytic glycoprotein 1 (PGP-1), Phagocytic glycoprotein I (PGP-I), CD44
About This Item
biological source
rat
Quality Level
conjugate
violetFluor® 450
antibody form
purified antibody
antibody product type
primary antibodies
clone
IM7, monoclonal
species reactivity
mouse, human
packaging
antibody small pack of 25 μg
technique(s)
flow cytometry: suitable
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... CD44(960)
mouse ... Cd44(12505)
General description
Immunogen
Application
Inflammation & Immunology
Immunoglobulins & Immunology
Quality
Flow Cytometry Analysis: 0.25 μg from a representative lot detected CD44, in one million C57Bl/6 splenocytes.
Physical form
Storage and Stability
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service