Skip to Content
Merck
All Photos(1)

Key Documents

MABF150C

Sigma-Aldrich

Anti-IL-2 (mouse), APC, clone JES6-5H4 Antibody

clone JES6-5H4, from rat, Allophycocyanin conjugate

Synonym(s):

Interleukin-2 (IL-2), T-cell growth factor (TCGF), IL-2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$189.00
100 μG
$411.00

$189.00


Estimated to ship on21 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$189.00
100 μG
$411.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$189.00


Estimated to ship on21 April 2025


Request a Bulk Order

biological source

rat

Quality Level

conjugate

Allophycocyanin conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

JES6-5H4, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Il2(16183)

General description

The JES6-5H4 antibody binds to mouse Interleukin-2 (IL-2), a 17 kDa cytokine that is secreted by activated T cells. This cytokine acts as an autocrine stimulatory factor for T cell proliferation, differentiation and survival, as well as being an effective activator of B cells and inducer of NK cell cytotoxic functions. IL-2 plays a key modulatory role in the differentiation of T cells toward either Th17 or T regulatory (Treg) cell types and is important for Treg cell function. The IL-2 receptor (IL-2R) is a complex consisting of an IL-2R alpha chain (CD25), along with the IL-2R beta chain (CD122) and the common gamma chain (CD132). IL-2 may also bind with low affinity to the T cell surface protein CD25 alone, although this interaction does not induce cell signaling.

Application

This rat monoclonal Anti-IL-2 Antibody (mouse), APC, clone JES6-5H4, Cat. No. MABF1505 is validated for use in Flow Cytometry, for the detection of IL-2.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.06 μg from a representative lot detected IL-2 in one million C57Bl/6 splenocytes.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service