Skip to Content
Merck
All Photos(1)

Key Documents

MABF1466

Sigma-Aldrich

Anti-CD45 (Mouse), clone 30-F11 Antibody

clone 30-F11, from rat

Synonym(s):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen (L-CA), Lymphocyte antigen 5 (Ly-5), T200, CD45, CD45

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$190.00

$190.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$190.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$190.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

30-F11, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

Related Categories

General description

The 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others. The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This rat monoclonal Anti-CD45 Antibody (Mouse), clone 30-F11, Cat. No. MABF1466 is validated for use in Flow Cytometry, for the detection of CD45.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected CD45 in one million C57Bl/6 splenocytes.

Physical form

Affinity Chromatography
Format: Purified
Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service