Skip to Content
Merck
All Photos(1)

Key Documents

MABF1418

Sigma-Aldrich

Anti-TIGIT (mouse), PE, clone 1G9 Antibody

clone 1G9, from mouse, PE

Synonym(s):

T-cell immunoreceptor with Ig and ITIM domains, V-set and transmembrane domain-containing protein 3, TIGIT

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$189.00

$189.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$189.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$189.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G9, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

The 1G9 antibody reacts with mouse TIGIT (T cell Ig and ITIM domain), a 26 kDa member of the CD28 receptor family which is reported to regulate T cell receptor (TCR) activation. Within the CD28 family of receptors there are those which have co-stimulatory activity, such as CD28 and CTLA-4, as well as more recently identified receptors like TIGIT which are proposed to provide co-inhibitory signals. TIGIT is expressed and upregulated on activated T cells, and is also expressed on memory and regulatory T cells. Upon engagement by its ligands, CD112 and CD155, TIGIT signaling inhibits T cell proliferation and suppresses T cell responses, without triggering cell deletion. A second inhibitory effect of TIGIT signaling is the generation of immunoregulatory dendritic cells, which secrete IL-10 and TGF-beta to further inhibit T cell function.The 1G9 antibody may be used for flow cytometric analysis of TIGIT, which is expressed at very high levels on T regulatory cells (Tregs) and activated conventional T cells, as well as memory T cells and NK cells.
Anti-TIGIT (mouse), PE, clone 1G9 Antibody is an antibody against TIGIT for use in Flow Cytometry.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.25 μg from a representative lot detected TIGIT in one million C57Bl/6 splenocytes.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service