Skip to Content
Merck
All Photos(1)

Key Documents

MABF1079Z

Sigma-Aldrich

Mouse IgG2b Negative Control, clone MPC-11, Azide Free Antibody

clone MPC-11, from mouse

Synonym(s):

Immunoglobulin G 2b, IgG2b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$190.00

$190.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$190.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$190.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

clone

MPC-11, monoclonal

technique(s)

flow cytometry: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

unmodified

General description

The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Application

This mouse monoclonal Mouse IgG2b Antibody Negative Control, clone MPC-11, Azide Free, Cat. No. MABF1079Z is validated for use in Flow Cytometry, for the detection of IgG2b.

Physical form

Format: Purified
Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, pH7.2. This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.

Other Notes

Concentration: Please refer to lot specific datasheet.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sangmin Lee et al.
PloS one, 14(2), e0212357-e0212357 (2019-02-16)
Antibodies, including antibodies to the RNA binding protein heterogeneous nuclear ribonucleoprotein A1, have been shown to contribute to the pathogenesis of multiple sclerosis, thus it is important to assess their biological activity using animal models of disease. Near-infrared optical imaging

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service