Skip to Content
Merck
All Photos(1)

Key Documents

MABE979

Sigma-Aldrich

Anti-Histone Demethylase JARID1C Antibody, clone 2B4.2

clone 2B4.2, from mouse

Synonym(s):

Lysine-specific demethylase 5C, Histone demethylase JARID1C, Jumonji/ARID domain-containing protein 1C, Protein SmcX, Protein Xe169

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$598.00

$598.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$598.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$598.00


Estimated to ship on10 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B4.2, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM5C(8242)
mouse ... Kdm5C(20591)

General description

Lysine-specific demethylase 5C (UniProt P41229; also known as Histone demethylase JARID1C, Jumonji/ARID domain-containing protein 1C, Protein SmcX, Protein Xe169) is encoded by the KDM5C (also known as DXS1272E, JARID1C, MRXSCJ, SMCX, XE169) gene (Gene ID 8242) in human. RBP2 (JARID1A), PLU-1 (JARID1B), KDM5C (JARID1C), and SMCY (JARID1D) constitute the Jumonji/ARID (JARID) subfamily of JmjC-containing proteins. KDM5C/JARID1C catalyzes the demethylation of di- and trimethylated lysine 4 of histone 3 in a Fe(II) and α-ketoglutarate-dependent manner. JARID1 catalytic activity is medidated by its JmjC domain (a.a. 468-634), while the JmjN domain (a.a. 14-55) and the C5HC2 zinc finger (a.a. 326-372 & 1187-1248) are both important in assisting JARID1C catalytic activity. JARID1C also contains an AT-rich domain-interacting domain (ARID) that binds DNA and its N-terminal PHD (plant homeodomain) finger (a.a. 326-372) is reported to bind trimethylated lysine 9 on histone 3 (H3K9me3). JARID1C interacts with PCNA through its PCNA-interaction protein (PIP) motif and JARID1C is among 485 chromatin factors enriched at nascent versus mature post-replicative chromatin, indicating a potential transcription-independent role during DNA replication.

Specificity

Clone 2B4.2 targets an N-terminal region sequence present in all human and murine JARID1C/KDM5C spliced isoforms reported by UniProt (P41229 and P41230).

Immunogen

Epitope: N-terminal region.
KLH-conjugated linear peptide corresponding to an N-terminal region sequence of human JARID1C.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
This Anti-Histone Demethylase JARID1C Antibody, clone 2B4.2 is validated for use in Western Blotting for the detection of Histone Demethylase JARID1C.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected histone demethylase JARID1C in 10 µg of HeLa cell lysate.

Target description

~180 kDa observed. 175.7/175.3 kDa (human/mouse isoform 1), 175.4/175.0 kDa (human/mouse isoform 2), 170.6/170.2 kDa (human/mouse isoform 3), 155.1/175.6 kDa (human isoform 4/5) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service