Skip to Content
Merck
All Photos(1)

Documents

MABE959

Sigma-Aldrich

Anti-MBNL2 Antibody, clone MB2a(3B4)

clone MB2a(3B4), 1 mg/mL, from mouse

Synonym(s):

Muscleblind-like protein 2, Muscleblind-like protein 1, Muscleblind-like protein-like, Muscleblind-like protein-like 39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MB2a(3B4), monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MBNL2(10150)

General description

MBNL2, also known as Muscleblind-like protein 2, Muscleblind-like protein 1, Muscleblind-like protein-like, or Muscleblind-like protein-like 39, and encoded by the gene MBNL2/MBLL/MBLL39/MLP1, is a protein that mediates pre-mRNA alternative splicing regulation. MBNL2 acts either as an activator or repressor of splicing on specific pre-mRNA targets. For instance MBNL2 inhibits cardiac troponin-T pre-mRNA exon inclusion but induces insulin receptor pre-mRNA exon inclusion in muscle. MBNL2 may also regulate ITGA3 expression by transporting it from the nucleus to cytoplasm at adhesion plaques. MBNL2 appears to play a key role in the muscle disease named myotonic dystrophy pathophysiology. MBNL2 is expressed in the nucleus of cells in multiple tissues but can be found in the cytoplasm in certain situations. In particular it is found in the cytoplasm in adhesion plaques.

Immunogen

Recombinant protein corresponding to human MBNL2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-MBNL2 Antibody, clone MB2a(3B4) is validated for use in Western Blotting for the detection of MBNL2.

Quality

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MBNL2 in 10 µg of C2C12 cell lysate.

Target description

~40 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service