Skip to Content
Merck
All Photos(1)

Key Documents

MABE915

Sigma-Aldrich

Anti-Mi-2β Antibody, clone 2G8

clone 2G8, from mouse

Synonym(s):

Chromodomain-helicase-DNA-binding protein 4, CHD-4, ATP-dependent helicase CHD4, Mi-2 autoantigen 218 kDa protein, Mi2-beta, Mi-2b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$599.00

$599.00


Estimated to ship on10 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$599.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$599.00


Estimated to ship on10 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G8, monoclonal

species reactivity

human

technique(s)

ChIP: suitable (ChIP-seq)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHD4(1108)

Related Categories

General description

Chromodomain-helicase-DNA-binding protein 4 (CHD-4), often referred to as Mi-2β, is a central component of the nucleosome remodeling and histone deacetylase (NuRD) repressor complex. Mi-2β has been shown to interact with TRIM27, HDAC1, Histone deacetylase 2, MTA2, SATB1 and Ataxia telangiectasia and Rad3 related. This protein also interacts with KLF1. Sumoylation of KLF1 leads to transcriptional repression of Mi-2β.

Application

Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody used as part of an antibody cocktail of consisting of Anti-Mi-2β clones 2G8, 16G4 and 17H11 (cat. no. MABE915, MABE914, and MABE920 respectively) was reported to work in ChIP (see Zhang, J., et al 2012).

ChIP-seq: A representative lot of this antibody used as part of an antibody cocktail of consisting of Anti-Mi-2β clones 2G8, 16G4 and 17H11 (cat. no. MABE915, MABE914, and MABE920 respectively) was reported to work in ChIP-seq (see Zhang, J., et al 2012).
This Anti-Mi-2β Antibody, clone 2G8 is validated for use in Western Blotting and ChIP-seq and Culture Confirmation for the detection of Mi-2β.

Quality

Evaluated by Western Blotting in a Jurkat cell lysate.

Western Blotting Analysis: A 1 µg/mL dilution of this antibody detected Mi-2β in 10 µg of a Jurkat cell lysate.

Target description

~ 220 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service