Skip to Content
Merck
All Photos(1)

Key Documents

MABE873

Sigma-Aldrich

Anti-Chd2, clone 8H3 Antibody

clone 8H3, from rat

Synonym(s):

SNF2 chromatin remodelling enzyme, Chromodomain Helicase DNA Binding Protein 2, Protein Chd2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$750.00

$750.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$750.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$750.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8H3, monoclonal

species reactivity

mouse, human

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHD2(1106)

General description

Chd2, also known as Chromodomain-helicase-DNA-binding protein 2 (CHD-2), or ATP-dependent helicase CHD2, and encoded by the gene CHD2, belongs to the CHD family of proteins. This family is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes are chromatin remodeling proteins and alter gene expression by modifying the chromatin structure of a particular gene. The alteration affects the access of the transcriptional apparatus to it, and thus the gene’s expression. In the mouse Chd2 is expressed in heart, forebrain, extremities, and facial and dorsal regions during specific times of embryonic development. Chd2 null mice are embryonic lethal, and in humans, various mutations in CHD-2 gene appear to cause a variety of disorders and developmental defects. Other mutations associated with the CHD2 gene locus appear to cause childhood-onset epileptic encephalopathy (EEOC).

Immunogen

Recombinant protein corresponding to mouse Chd2.

Application

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Chd2 from C2C12 cell lysate (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Chd2 in C2C12 cells (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Chd2 from C2C12 cell lysate (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-Chd2 antibody, clone 8H3 is validated for use in western blotting, IP, ICC & ChIP for the detection of Chd2.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Chd2 in 10 µg of Jurkat cell lysate.

Target description

~211 kDa observed. Uncharacterized band(s) may be observed in some cell lysates. Two known isoforms may be observed at 50 kDa and 200 kDa

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziqian Min et al.
iScience, 25(12), 105552-105552 (2022-11-30)
Chromodomain helicase DNA-binding domain 2 (CHD2) is a chromatin remodeling factor involved in many developmental processes. However, its role in male germ cell development has not been elucidated. Here, we confirm that CHD2 expression is enriched in the male germline.
Aviv Rom et al.
Nature communications, 10(1), 5092-5092 (2019-11-11)
Chromodomain helicase DNA binding protein 2 (Chd2) is a chromatin remodeller implicated in neurological disease. Here we show that Chaserr, a highly conserved long noncoding RNA transcribed from a region near the transcription start site of Chd2 and on the
Alberto Parras et al.
Nature, 560(7719), 441-446 (2018-08-17)
Common genetic contributions to autism spectrum disorder (ASD) reside in risk gene variants that individually have minimal effect sizes. As environmental factors that perturb neurodevelopment also underlie idiopathic ASD, it is crucial to identify altered regulators that can orchestrate multiple

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service