Skip to Content
Merck
All Photos(2)

Key Documents

MABE645

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys9) Antibody, clone RM161

clone RM161, from rabbit

Synonym(s):

Histone H3.3, Histone H3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$711.00

$711.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$711.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$711.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RM161, monoclonal

species reactivity

human

technique(s)

multiplexing: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys9)

Gene Information

human ... H3F3B(3021)

General description

Histone H3, also known as Histone H3.1t (H3/t), H3t, H3/g, and encoded by the gene name HIST3H3/ H3FT, is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Therefore, histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. High levels of phosphorylation of Histone H3 are associated with mitosis.

Specificity

This antibody reacts to Histone H3 acetylated at Lysine 9 (K9ac). No cross reactivity with other acetylated Lysines in Histone H3.

Immunogen

A linear peptide corresponding to human Histone H3 acetylated at Lys9.

Application

This Anti-acetyl-Histone H3 (Lys9) Antibody, clone RM161 is validated for use in Western Blotting, Multiplexing for the detection of acetyl-Histone H3.

Quality

Evaluated by Western Blotting in untreated and sodium butyrate treated HeLa acid extract.

Western Blotting Analysis (WB): 0.25 μg/mL of this antibody detected acetyl-Histone H3 (Lys9) in sodium butyrate treated HeLa acid extract.

Target description

~17 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William J Lu-Culligan et al.
Nature, 622(7981), 173-179 (2023-09-21)
Lysine residues in histones and other proteins can be modified by post-translational modifications that encode regulatory information1. Lysine acetylation and methylation are especially important for regulating chromatin and gene expression2-4. Pathways involving these post-translational modifications are targets for clinically approved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service