Skip to Content
Merck
All Photos(2)

Key Documents

MABE636

Sigma-Aldrich

Anti-monomethyl Histone H3 (Lys4) Antibody, clone RM140

clone RM140, from rabbit

Synonym(s):

Histone H3.3, Histone H3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$656.00

$656.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$656.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$656.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

RM140, monoclonal

species reactivity

human

technique(s)

multiplexing: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys4)

Gene Information

human ... H3F3B(3021)

General description

Histone H3, also known as Histone H3.1t (H3/t), H3t, H3/g, and encoded by the gene name HIST3H3/ H3FT, is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Therefore, histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. High levels of phosphorylation of Histone H3 are associated with mitosis.

Specificity

This antibody reacts to Histone H3 monomethylated at Lysine 4 (K4me1). No cross reactivity with dimethylated Lysine 4 (K4me2) or trimethylated Lysine 4 (K4me3), or other methylations in Histone H3.

Immunogen

A linear peptide corresponding to human Histone H3 monomethylated at Lys4.

Application

Multiplex Analysis (Mplex): 0.1-0.5 μg/mL from a representative lot was used in Multiplex.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
This Anti-monomethyl Histone H3 (Lys4) Antibody, clone RM140 is validated for use in Western Blotting, Multiplexing for the detection of monomethyl Histone H3.

Quality

Evaluated by Western Blotting in Recombinant HeLa protein and HeLa acid extract.

Western Blotting Analysis (WB): 0.5 μg/mL of this antibody detected monomethyl Histone H3 (Lys4) in HeLa acid extract.

Target description

~17 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jimsheena V Karakkat et al.
Nucleic acids research, 47(19), 10086-10103 (2019-09-19)
The metabolic sensor Per-Arnt-Sim (Pas) domain-containing serine/threonine kinase (PASK) is expressed predominantly in the cytoplasm of different cell types, although a small percentage is also expressed in the nucleus. Herein, we show that the nuclear PASK associates with the mammalian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service