Skip to Content
Merck
All Photos(2)

Key Documents

MABE432

Sigma-Aldrich

Anti-TRIM33 Antibody, clone 3B5.1

clone 3B5.1, 1 mg/mL, from mouse

Synonym(s):

E3 ubiquitin-protein ligase TRIM33, Ectodermin homolog, RET-fused gene 7 protein, Protein Rfg7, Transcription intermediary factor 1-gamma, TIF1-gamma, Tripartite motif-containing protein 33

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$350.00

$350.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$350.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$350.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B5.1, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRIM33(51592)

General description

TRIM33, also known as E3 ubiquitin-protein ligase TRIM33, Ectodermin homolog, RET-fused gene 7 protein, Rfg7, Transcription intermediary factor 1-gamma, TIF1-gamma, or Tripartite motif-containing protein 33, and encoded by the gene TRIM33/KIAA1113/RFG7/TIF1G, is an E3 ubiquitin protein ligase. Among its targets is SMAD4 for which ubiquitination facilitates its nuclear exclusion and degradation. SMAD3 is also a target for ubiquitination, and together, TRIM33′s association promotes erythroid differentiation of hematopoietic stem cells. Finally mono-ubiquitination of various targets by TRIM33 affects their ability to form complexes which often hampers their activity and thus regulates their subsequent actions. Such is the case of SMAD4 and its interaction with TGF-beta/BMP signaling. Monoubiquitination of SMAD4 by TRIM33 hampers its ability to form a stable complex with activated SMAD2/3 resulting in inhibition of TGF-beta/BMP signaling cascade. TRIM33 is localized in nucleus in nuclear dots or bodies and is expressed in most tissues and stem cells during development as well as in the adult in multiple tissues.

Immunogen

GST-tagged recombinant protein corresponding to human TRIM33.

Application

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected TRIM33 in H9 stem cells.
Alexa Fluor is a registered trademark of Life Technologies.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-TRIM33 Antibody, clone 3B5.1 is validated for use in Western Blotting and Immunocytochemistry for the detection of TRIM33.

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected TRIM33 in 10 µg of K562 cell lysate.

Target description

~123 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service