Skip to Content
Merck
All Photos(1)

Key Documents

MABE1820

Sigma-Aldrich

Anti-Neurofibromin C Antibody, clone NFC

clone NFC, from mouse

Synonym(s):

Neurofibromatosis-related protein NF-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$252.00
100 μL
$769.00

$252.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$252.00
100 μL
$769.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NFC, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NF1(4763)

General description

Neurofibromin (UniProt: P21359; also known as Neurofibromatosis-related protein NF-1) is encoded by the NF1 gene (Gene ID: 4763) in human. Neurofibromin is a GTPase-activating protein that negatively regulatres RAS/MAOK pathway by enhancing the hydrolysis of Ras-bound GTP. Hence, it can display tumor suppressor properties. Neurofibromin has been detected in brain, peripheral nerves, lung, colon, and muscle tissues. It has a CRAL-TRIO domain (aa 1580-1738) that facilitates its binding to phospholipids. It binds primarily glycerophospholipids with monounsaturated C18:1 and/or C16:1 fatty acid moieties and a phosphatidylethanolamine or phosphatidylcholine head group. Neurofibromin also has a Ras-GTP domain (aa 1235-1451). Mutations in NF1 gene are known to cause Neurofibromatosis type I that involved tumors in the peripheral nervous system and fibromatous skin tumors. Some mutations are also known to cause colorectal cancer and juvenile leukemia. Six different isoforms of Neurofibromin have been described that are produced by alternative splicing. (Ref.: Rossi, S., et al. (2018). Mod. Pathol. 31(1), 160-168).

Specificity

Clone NFC specifically detects human Neurofibromin C. it targets an epitope with in the C-terminal region.

Immunogen

Epitope: C-terminus
Recombinant fragment corresponding to 281 amino acids from the C-terminal region of human Neurofibromin C.

Application

Anti-Neurofibromin C, clone NFC, Cat. No. MABE1820, is a highly specific mouse monoclonal antibody that targets Neurofibromin and has been tested for use in Immunohistochemistry (Paraffin).
Immunohistochemistry (Paraffin) Analysis: A representative lot detected Neurofibromin C in Immunohistochemistry applications (Rossi, S., et. al. (2018). Mod Pathol. 31(1):160-168).
Western Blotting Analysis: A representative lot detected Neurofibromin C lysate from human NF1 +/+ cells, but not in -/- cells or NF1+/+ cells transfected with siRNA targeting neurofibromin (Reuss, DE.,et al., (2014). Acta Neuropathol. 127(4); 565-572).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Immunohistochemistry (Paraffin) in human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Neurofibromin C in human kidney tissue sections.

Target description

319.37 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jocelyne Lopez et al.
The Journal of biological chemistry, 299(6), 104789-104789 (2023-05-07)
Sprouty-related EVH-1 domain-containing (SPRED) proteins are a family of proteins that negatively regulate the RAS-Mitogen-Activated Protein Kinase (MAPK) pathway, which is involved in the regulation of the mitogenic response and cell proliferation. However, the mechanism by which these proteins affect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service