Skip to Content
Merck
All Photos(1)

Key Documents

MABE1144

Sigma-Aldrich

Anti-TET1 Antibody, clone 5D6

clone 5D6, from rat

Synonym(s):

Methylcytosine dioxygenase TET1, CXXC-type zinc finger protein 6, Ten-eleven translocation 1 gene protein homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$598.00

$598.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$598.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$598.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D6, monoclonal

species reactivity

mouse

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Tet1(52463)

General description

Methylcytosine dioxygenase TET1 (EC 1.14.11.n2; UniProt Q3URK3; also known as CXXC-type zinc finger protein 6, Ten-eleven translocation 1 gene protein homolog) is encoded by the Tet1 (also know as Cxxc6, Kiaa1676) gene (Gene ID 52463) in murine species. TET (ten-eleven translocation) proteins (TET1, TET2, and TET3) oxidize 5-methylcytosine to 5-hydroxymethylcytosine, 5-formylcytosine, and 5-carboxylcytosine, providing a means for active epigenetic DNA demethylation in mammals. TET proteins also modify genomic thymine residues to 5-hydroxyuracil. TET proteins depend on Fe(II) and 2-oxoglutarate as cofactors for activity, and 2-oxoglutarate is known to inhibit their activity, while ascorbate is shown to stimulate TET-mediated cytosine oxidation. In addition, post-translational modifications (PTMs), such as phosphorylation and O-GlcNAcylation, also play a role in regulating the activity of TET proteins. TET1 and TET2 are found highly expressed in mouse embryonic stem cells (mESCs) and TET3 is known to be upregulated in oocytes and oxidize the silenced paternal pronuclear DNA. High levels of TET proteins and genomic 5-hydroxymethylcytosine in neuronal tissues are also reported.

Specificity

Clone 5D6 detected exogenously expressed TET1, but not TET2 or TET3, GST fusion protein (Bauer, C., et al. (2015). J. Biol. Chem. 290(8):4801-4812).

Immunogen

Epitope: Region between DNA-binding and 2-oxoglutarate-binding domains
His-tagged recombinant fragment from the region between DNA-binding and 2-oxoglutarate-binding domains of mouse TET1.

Application

Anti-TET1 Antibody, clone 5D6 is an antibody against TET1 for use in Immunocytochemistry, Western Blotting, Immunoprecipitation, ELISA.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blotting Analysis: A representative lot detected the endogenous TET1 in murine embryonic stem cell (mESC) lysate, as well as exogenously expressed TET1, but not TET2 or TET3, GST fusion in lysates from respective HEK293T transfectants (Bauer, C., et al. (2015). J. Biol. Chem. 290(8):4801-4812).
Immunoprecipitation Analysis: A representative lot immunoprecipitated endogenous TET1 from murine embryonic stem cell (mESC) lysate. Subsequent LC-MS/MS analysis detected the presence of co-immunoprecipitated glycosyltransferase OGT (Bauer, C., et al. (2015). J. Biol. Chem. 290(8):4801-4812).
ELISA Anaylsis: Clone 5D6 hybridoma culture supernatant was confirmed for its immunoreactivity against His-tagged murine TET1 by ELISA (Bauer, C., et al. (2015). J. Biol. Chem. 290(8):4801-4812).

Quality

Evaluated by Immunocytochemistry in mouse ESCs.

Immunocytochemistry Analysis: A 1:200 dilution of this antibody detected TET1 in mouse embryonic stem cells (ESCs).

Target description

219.3 calculated.

Linkage

Replaces: ABE1034

Physical form

Format: Purified
Protein G purified.
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linlin Liu et al.
Frontiers in cell and developmental biology, 9, 644135-644135 (2021-04-10)
Tet enzymes participate in DNA demethylation and play critical roles in stem cell pluripotency and differentiation. DNA methylation alters with age. We find that Tet1 deficiency reduces fertility and leads to accelerated reproductive failure with age. Noticeably, Tet1-deficient mice at
Benedict Shi Xiang Lian et al.
iScience, 25(4), 104118-104118 (2022-04-12)
The innate immune system is an immediate defense against infectious pathogens by the production of inflammatory cytokines and other mediators. Deficiencies of epigenetic regulatory enzymes, such as Tet1 and Dnmt1, cause dysregulation of cytokine expression. However, it is unclear if

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service