Skip to Content
Merck
All Photos(3)

Key Documents

MABE1135

Sigma-Aldrich

Anti-TRF2 Antibody, clone T2JKM3

clone T2JKM3, from mouse

Synonym(s):

Telomeric repeat-binding factor 2, TTAGGG repeat-binding factor 2, Telomeric DNA-binding protein, TERF2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$252.00
100 μG
$641.00

$252.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$252.00
100 μG
$641.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

T2JKM3, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TERF2(7014)

General description

Telomeric repeat-binding factor 2 (UniProt: Q15554; also known as TTAGGG repeat-binding factor 2, Telomeric DNA-binding protein, TERF2) is encoded by the TERF2 (also known as TRBF2, TRF2) gene (Gene ID: 7014) in human. TERF2 is homodimeric protein that is ubiquitous in its expression. Higher expression levels are reported in spleen, thymus, prostate, uterus, testis, peripheral blood leukocytes, colon, and small intestine. Two soforms of TERF2 are reported that are produced by alternative splicing.TERF2 binds to telomeric double-stranded 5′-TTAGGG-3′ repeat and plays a central role in telomere maintenance and protection against end-to-end fusion of chromosomes. It is also required to recruit a number of factors and enzymes required for telomere protection, including the sheltering complex, TERF2IP/RAP1 and DCLRE1B/Apollo. TEFR2 colocalizes with telomeric DNA in interphase cells and is located at chromosome ends during metaphase. It is phosphorylated upon DNA damage and the phosphorylated TERF2 is not bound to telomeric DNA, and rapidly localizes to damage sites. Inhibition of TRF2 causes apoptosis in several mammalian cell types and its over-expression is observed in a variety of human cancers.

Specificity

Clone T2JKM3 detects Telomeric repeat-binding factor 2 in human cells.

Immunogen

Tagged full length recombinant human Telomeric repeat-binding factor 2.

Application

Anti-TRF2, clone T2JKM3, Cat. No. MABE1135, is a highly specific mouse monoclonal antibody that targets Telomeric repeat-binding factor 2 (TRF2) and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected TRF2 in IMR90 primary human fibroblasts.

Western Blotting Analysis: A 1:200 dilution from a representative lot detected TRF2 in HT1080 cell lysate.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HT1080 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected TRF2 in HT1080 cell lysate.

Target description

~60 kDa observed; 59.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service