Skip to Content
Merck
All Photos(2)

Key Documents

MABE1130

Sigma-Aldrich

Anti-Hsp70/Hsp72 Antibody, clone 2H9

clone 2H9, from mouse

Synonym(s):

Heat shock 70 kDa protein 1A, Heat shock 70 kDa protein 1, HSP70.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$563.00

$563.00


Check Cart for Availability


Select a Size

Change View
100 μG
$563.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$563.00


Check Cart for Availability

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H9, monoclonal

species reactivity

bovine, human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HSPA1A(3303)

General description

Heat shock 70 kDa protein 1A (UniProt Q27975; also known as Heat shock 70 kDa protein 1, HSP70.1) is encoded by the HSPA1A (also known as HSP70-1) gene (Gene ID 281825 & 282254) in bovine species. The mammalian HSP70 family consists of the mitochondrial HSP75, the endoplasmic HSP78, the cytoplasmic HSC70 (a.k.a. HSC73 and HSP73) and its inducible counterpart HSP72 (a.k.a. HSP70). The latter two have similar structures and properties, and both of them bind misfolded, newly synthesized polypeptides in an ATP-dependent manner, reflecting their chaperonic activity. All cells constitutively express HSC70. On the other hand, the expression of HSP72 in normal cells is mainly stress inducible, although constitutively high HSP72 expressions are observed in malignant human tumors of various origins. The induced HSP72 expression enables stressed cells to better cope with the elevation of unfolded/denatured proteins and to prevent apoptosis under various stress conditions.

Specificity

Clone 2H9 reacts with the heat schock-inducible HSP72 (HSPA1A), but not the constitutively expressed HSPA8/Hsc70 (HSC73) (Demidov, O.N., et al. (1999). Eur. J. Cardiothorac. Surg. 16(4):444-449). Reactivity toward HSPA1B and HSPA2 (UniProt Q27965 and P34933) has not been determined.

Immunogen

Bovine skeletal muscle Hsp70/Hsp72.

Application

Anti-Hsp70/Hsp72 Antibody, clone 2H9 is an antibody against Hsp70/Hsp72 for use in Western Blotting.
Immunoprecipitation Analysis: Clone 2H9 ascites fluid immunoprecipitated Hsp70/Hsp72 from human U-937 myeloid leukemia cell lysates (Courtesy of Dr. Irina V. Guzhova and Dr. Boris Margulis, Russian Academy of Sciences).
Western Blotting Analysis: A representative lot detected Hsp70/Hsp72 expression and siRNA-mediated Hsp70 down-regulation in HeLa cells (Rohde, M., et al. (2005). Genes Dev. 19(5):570-582).
Western Blotting Analysis: A representative lot detected the heat schock-inducible Hsp70/Hsp72, but not the constitutively expressed HSPA8/Hsc70 (HSC73) in heat shock protein preparations from bovine skeletal muscle and human myocardium tissue (Demidov, O.N., et al. (1999). Eur. J. Cardiothorac. Surg. 16(4):444-449).
Western Blotting Analysis: A representative lot detected heat shock-induced Hsp70 expression in four human leukemia cell lines (U937, Jurkat, Molt4, HL60), but not in murine myeloma NSO cells (Lasunskaia, E.B., et al. (1997). Apoptosis. 2(2):156-163).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chaperones

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Hsp70/Hsp72 in 10 µg of MCF7 cell lysate.

Target description

~70 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasiia Griukova et al.
Aging, 11(21), 9912-9931 (2019-11-07)
Hormone-regulated proliferation and differentiation of endometrial stromal cells (ESCs) determine overall endometrial plasticity and receptivity to embryos. Previously we revealed that ESCs may undergo premature senescence, accompanied by proliferation loss and various intracellular alterations. Here we focused on whether and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service