Skip to Content
Merck
All Photos(3)

Key Documents

MABE1116

Sigma-Aldrich

Anti-DNAJB1/Hdj1 Antibody, clone J25

clone J25, from mouse

Synonym(s):

Hdj1, DnaJ homolog subfamily B member 1, DNAJB1, hDj-1, Heat shock 40 kDa protein 1, Heat shock protein 40, HSP40

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$485.00

$485.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$485.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$485.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

J25, monoclonal

species reactivity

rat, mouse, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DNAJB1(3337)

General description

DnaJ homolog subfamily B member 1 (UniProt Q3MI00) is encoded by the DNAJB1 gene (Gene ID 538426) in bovine species. Heat shock proteins (Hsps) are classified into six major families based on their relative molecular mass, Hsp110/hspH, Hsp90/HspC, Hsp70/HspA, Hsp60/HspD, Hsp40/DnaJ, and small heat shock proteins sHsp/HspB. Hsp40/DnaJB1 belongs to the Hsp40/DnaJ subfamily of heat shock proteins. Hsp40 acts as a co-chaperone with Hsp70 to facilitate protein folding, transport, and degradation. MK5 is reported to stimulate the ATP hydrolyse activity of Hsp40/Hsp70 complex by phosphorylating Hsp40/DnaJB1 C-terminal serine residues Ser149, Ser151, Ser171, resulting in an enhancement of Hsp40/DnaJB1-dependent suppression of heat shock factor 1-driven transcription. DnaJB1 is also shown to interact with MDM2 and prevent MDM2-mediated ubiquitination and proteasomal degradation of p53. DnaJB1 depletion promotes cancer cell growth as a result of p53 pathway inactivation.

Immunogen

Purified bovine DNAJB1.

Application

Immunocytochemistry Analysis: 1-5 µg/mL from a representative lot detected heat shock-induced DNAJB1 upreguation in C6 rat glioma, B16 mouse melanoma, and K-562 human erythroleukemia cells (Courtesy of Dr. Irina V. Guzhova and Dr. Boris Margulis, Russian Academy of Sciences).
Flow Cytometry Analysis: 1-5 µg/mL from a representative lot detected heat shock-induced DNAJB1 upreguation in C6 rat glioma, B16 mouse melanoma, and K-562 human erythroleukemia cells (Courtesy of Dr. Irina V. Guzhova and Dr. Boris Margulis, Russian Academy of Sciences).
Western Blotting Analysis: 0.05-0.5 µg/mL from a representative lot detected heat shock-induced DNAJB1 upreguation in C6 rat glioma, B16 mouse melanoma, and K-562 human erythroleukemia cells (Courtesy of Dr. Irina V. Guzhova and Dr. Boris Margulis, Russian Academy of Sciences).
This Anti-DNAJB1/Hdj1 Antibody, clone J25 is validated for use in Western Blotting, Immunocytochemistry and Flow Cytometry for the detection of DNAJB1/Hdj1.

Quality

Evaluated by Western Blotting in human K562 erythroblastoma cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected heat shock-induced DNAJB1/Hdj-1 upregulation in human K-562 erythroleukemia cells.

Target description

~38 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service