Skip to Content
Merck
All Photos(1)

Key Documents

MABE1060

Sigma-Aldrich

Anti-CtIP Antibody, clone 14-1

clone 14-1, from mouse

Synonym(s):

CtBP-interacting protein, CtIP, Retinoblastoma-binding protein 8, RBBP-8, Retinoblastoma-interacting protein and myosin-like, RIM, Sporulation in the absence of SPO11 protein 2 homolog, SAE2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$616.00

$616.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$616.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$616.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14-1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RBBP8(5932)

General description

DNA endonuclease RBBP8 (UniProt: Q99708; also known CtBP-interacting protein, CtIP, Retinoblastoma-binding protein 8, RBBP-8, Retinoblastoma-interacting protein and myosin-like, RIM, Sporulation in the absence of SPO11 protein 2 homolog, SAE2) is encoded by the RBBP8 (also known as CTIP) gene (Gene ID: 5932) in human. RBBP-8 is an endonuclease that cooperates with the MRE11-RAD50-NBN (MRN) complex in processing meiotic and mitotic double-strand breaks (DSBs) by ensuring both resection and intrachromosomal association of the broken ends. RBBP8 is phosphorylated upon DNA damage (at Thr859) by ATR and this phosphorylated form binds to chromatin to initiate resection. The non-phosphorylated form does not bind to chromatin. It is reported that ATM activity is required for an early step in resection, leading to ATR activation, CtIP- phosphorylation, and accumulation of CtIP on chromatin. RBBP8 functions downstream of the MRN complex and promotes ATR activation and its recruitment to DSBs in the S/G2 phase facilitating the generation of ssDNA. It is localized predominantly in the nucleus and interacts with CTBP, with the C-terminal (BRCT) domains of BRCA1, and with the retinoblastoma protein. By interacting with BCRA1 it modulates transcriptional regulation, DNA repair, and cell cycle checkpoint control. Mutations in RBBP8 gene are known to cause Seckel syndrome 2, an autosomal recessive disorder characterized by proportionate dwarfism of prenatal onset associated with low birth weight, growth retardation, severe microcephaly, and mental retardation.

Specificity

Clone 14-1 specifically detects CtBP-interacting protein in HEK293 cells.

Immunogen

Epitope: domain of:
GST-tagged recombinant fragment corresponding to 278 amino acids from the C-terminal half of human CtBP-interacting protein.

Application

Anti-CtIP Antibody, clone 14-1, Cat. No. MABE1060, is a highly specific mouse monoclonal antibody that targets CtIP and has been used in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected CtIP in Immunocytochemistry applications (Murina, O., et. al. (2014). Cell Rep. 7(4):1030-8).

Western Blotting Analysis: A representative lot detected CtIP in Western Blotting applications (Yu, X., et. al. (2000). J Biol Chem. 275(24):18541-9; Bunting, S.F., et. al. (2010). Cell. 141(2):243-54; Wu, M., et. al. (2007). Mol Cancer Res. 5(12):1285-95).

Immunoprecipitation Analysis: A representative lot was used for CtIP Immunoprecipitation. (Yu, X., et. al. (2000). J Biol Chem. 275(24):18541-49).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.


Western Blotting Analysis: 0.5 µg/mL of this antibody detected CtIP in 10 µg of HEK293 cell lysate.


Target description

~120 kDa observed; 101.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John J Krais et al.
Nature communications, 12(1), 5016-5016 (2021-08-20)
DNA damage prompts a diverse range of alterations to the chromatin landscape. The RNF168 E3 ubiquitin ligase catalyzes the mono-ubiquitination of histone H2A at lysine (K)13/15 (mUb-H2A), forming a binding module for DNA repair proteins. BRCA1 promotes homologous recombination (HR)
Priya Kapoor-Vazirani et al.
Nature communications, 13(1), 6707-6707 (2022-11-08)
Sterile alpha motif and HD domain-containing protein 1 (SAMHD1) has a dNTPase-independent function in promoting DNA end resection to facilitate DNA double-strand break (DSB) repair by homologous recombination (HR); however, it is not known if upstream signaling events govern this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service