Skip to Content
Merck
All Photos(2)

Key Documents

MABE1055

Sigma-Aldrich

Anti-RecQ1 Antibody, clone 5A12.1

clone 5A12.1, from mouse

Synonym(s):

ATP-dependent DNA helicase Q1, EC:3.6.4.12, DNA helicase, RecQ-like type 1, RecQ1, DNA-dependent ATPase Q1, RecQ protein-like 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$189.00
100 μL
$481.00

$189.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$189.00
100 μL
$481.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$189.00


Estimated to ship on23 April 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A12.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RECQL(5965)

General description

ATP-dependent DNA helicase Q1 (UniProt: P46063; also known as EC:3.6.4.12, DNA helicase, RecQ-like type 1, RecQ1, DNA-dependent ATPase Q1, RecQ protein-like 1) is encoded by the RECQL (also known as RECQ1, RECQL1) gene (Gene ID: 5965) in human. RecQ helicases are a ubiquitous family of DNA unwinding enzymes involved in the maintenance of chromosome stability. They participate in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Five members of the RecQ family have been reported in human cells: BLM, RECQ1, RECQ4, RECQ5, and WRN. RecQ1 displays high expression in heart, lung, skeletal muscle and kidney. It is also highly expressed in tumor cells and although brain exhibits low level of EecQ1, its expression is dramatically increased in human glioblastoma. Depletion of RecQ1 affects proliferation of glioblastoma cells and causes an increase in the level of DNA damage. RecQ1 depleted tumor cells are shown to be more sensitive to anti-tumor agents, such as hydroxyurea and temozolomide.

Specificity

Clone 5A12.1 specifically detects human ATP-dependent DNA helicase Q1 (RecQ1). It targets an epitope within 87 amino acids from the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 87 amino acids from the internal region of human ATP-dependent DNA helicase Q1.

Application

Anti-RecQ1, clone 5A12.1, Cat. No. MABE1055, is a highly specific mouse monoclonal antibody that targets RecQ1 and has been tested for use in Western Blotting and Immunohistochemistry (Paraffin).
Immunohistochemistry (Paraffin) Analysis: A 1:50-250 dilution from a representative lot detected RecQ1 in human kidney and human tonsil tissue sections.

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RecQ1 in Raji cell lysate.

Target description

~73 kDa observed; 73.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service