Skip to Content
Merck
All Photos(1)

Key Documents

MABE1039

Sigma-Aldrich

Anti-Nup96 Antibody, clone 4H5

clone 4H5, from rat

Synonym(s):

Nuclear pore complex protein Nup98-Nup96, Nuclear pore complex protein Nup98, 98 kDa nucleoporin, Nucleoporin Nup98, Nup98, Nuclear pore complex protein Nup96, 96 kDa nucleoporin, Nucleoporin Nup96, Nup96, Nup96

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$610.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4H5, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nup98(269966)

General description

The murine gene Nup98 (Gene ID 269966; also known as AI849286, 4732457F17) encodes the 1816-amino acid nuclear pore complex protein Nup98-Nup96 (UniProt Q6PFD9), which is posttranslationally autocleaved between aa 800 and 801 to form Nup98 (aa 1-800) and Nup96 (aa 801-1816). The nuclear pore complex (NPC) acts as a passageway for nucleocytoplasmic transport. The core structure of the NPC is maintained by the Nup93-205 complex sandwiched between the supporting structures of the Nup107-160 complex on the nuclear and cytoplasmic faces of the pore. Nup96 is a component of the Nup107-160 complex, the largest subunit of the NPC. Nup96 knockout is reported to be fatal in mice and Nup96(+/-) mice with low levels of Nup96 display impaired immune response and are highly susceptible to viral infection.

Specificity

Reacts with Nup98-Nup96 precursor (isoforms 1, 2, and 6), nucleoporin Nup96 (isoform 5), but not nucleoporin Nup98 (isoform 3).

Immunogen

Epitope: Near N-terminus.
GST-tagged recombinant protein corresponding to mouse Nup96 near the N-terminus.

Application

Anti-Nup96 Antibody, clone 4H5 is an antibody against Nup96 for use in Western Blotting and Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: A representative lot detected Nup96 in NIH/3T3 cells (Mizuguchi, C., et al. (2010). Hybridoma. 29(6):551-553).
Immunocytochemistry Analysis: A representative lot detected Nup96 in NIH/3T3 cells (Mizuguchi, C., et al. (2010). Hybridoma. 29(6):551-553).

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Nup96 in 10 µg of NIH/3T3 cell lysate.

Target description

~120 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tadashi Makio et al.
Molecular biology of the cell, 35(5), ar62-ar62 (2024-03-20)
The Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) accessory protein Orf6 works as an interferon antagonist, in part, by inhibiting the nuclear import activated p-STAT1, an activator of interferon-stimulated genes, and the export of the poly(A) RNA. Insight into the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service