Skip to Content
Merck
All Photos(3)

Key Documents

MABD151

Sigma-Aldrich

Anti-podocalyxin associated keratan sulfate, clone R-10G| MABD151 Antibody

clone R-10G, from mouse

Synonym(s):

Podocalyxin, PC, PCLP-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$751.00

$751.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$751.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$751.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

R-10G, monoclonal

species reactivity

human

should not react with

mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1, kappa

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PODXL(5420)

General description

Podocalyxin-like protein 1 is a sialomucin that is involved in the regulation of both cell adhesion and cell morphology through its interactions with several intracellular and at least one extracellular ligand. It is known to play an essential role in the formation and maintenance of podocyte foot processes, in cancer development and aggressiveness and as a marker of undifferentiatted embryonic stem cells. MABD151, a monoclonal antibody generated against human iPS cells, is a novel marker antibody recognizing a type of keratan sulfate lacking oversulfated structure on hiPS cells. MABD151 is a useful molecular probe for disclosing the roles of glycans on the surface of hiPS cells in the maintenance of self-renewal and pluripotency and also as a potent tool for the evaluation and standardization of hiPS cells with different tissue origins and different histories in regenerative medicine (Kawabe K et.al, (2013) Glycobiology. v23, 322–336.

Immunogen

Whole Human iPS cells (Tic cell line).

Application

Evaluated by Western Blot in human brain lysate.
Western Blot Analysis: 1µg/mL of this antibody detected podocalyxin associated keratin sulphate in 10 µg of human brain lysate.

Evaluated by Immunocytochemistry in human ES cells.
Immunocytochemical Analysis: A 1:250 dilution of this antibody detected odocalyxin associated keratin sulphate in human ES cells.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Anti-podocalyxin associated keratan sulfate antibody, clone R-10G is validated for use in ICC & western blotting for the detection of podocalyxin associated keratan sulfate.

Quality

Evaluated by Western Blot in human ES cell lysate.

Western Blot Analysis: 1µg/mL of this antibody detected podocalyxin associated keratin sulphate in 10 µg of human ES cell lysate.

Target description

above 250kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service