Skip to Content
Merck
All Photos(1)

Key Documents

MABD122A4

Sigma-Aldrich

Anti-CD73, clone AA60-E3-3, Alexa Fluor 488 conjugate Antibody

clone AA60-E3-3, from mouse, ALEXA FLUOR 488

Synonym(s):

CD-73, 5′-nucleotidase, 5′-NT, ecto-5′-nucleotidase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$514.00

$514.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$514.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

$514.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AA60-E3-3, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (immunogen homology)

technique(s)

immunocytochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NT5E(4907)

General description

CD73, also known as Ecto-5′-nucleotidase, catalyzes the conversion of purine 5′ mononucleotides to nucleosides, the preferred substrate being AMP. The enzyme consists of a dimer of 2 identical 70-kD subunits bound by a glycosyl phosphatidyl inositol linkage to the external face of the plasma membrane. The enzyme is used as a marker of lymphocyte differentiation, being expressed on subsets of B-cells and T-cells. CD73 is also a marker of undifferentiated mesenchymal stem cells and is often used to identified stem cell populations.

Immunogen

Whole Mouse Dermal Lymphatic Endothelial Cells (DLEC)

Application

Evaluated by Immunocytochemistry on human bone marrow mesenchymal stem cells. Immunocytochemical analysis: A 1:50 dilutions of this antibody detected CD73 on 1X10^6 human adipose and bone marrow mesenchymal stem cells.
Research Category
Stem Cell Research
Research Sub Category
Mesenchymal Stem Cells
This Anti-CD73, clone AA60-E3-3 is validated for use in Immunocytochemistry for the detection of CD73.

Quality

Evaluated by Immunocytochemistry on human adipose mesenchymal stem cells.
Immunocytochemical analysis: A 1:50 dilutions of this antibody detected CD73 on 1X10^6 human adipose and bone marrow mesenchymal stem cells.

Target description

The unconjugated parent antibody (Catalog No. MABD122) has an observed MW at 63 kDa

Physical form

Protein G Purified
Purified mouse monoclonal IgG1κ conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15mg/ml BSA.

Storage and Stability

Maintain refrigerated at 2-8ºC protected from light in undiluted aliquots for up to 6 months from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service