Skip to Content
Merck
All Photos(1)

Documents

MABC967

Sigma-Aldrich

Anti-Kallikrein 7/KLK7 Antibody, clone 73-1

clone 73-1, from mouse

Synonym(s):

KLK7, Kallikrein-7, hK7, Serine protease 6, Stratum corneum chymotryptic enzyme, hSCCE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

73-1, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... KLK7(5650)

General description

Kallikrein-7 (UniProt: P49862; also known as hK7, Serine protease 6, Stratum corneum chymotryptic enzyme, hSCCE) is encoded by the KLK7 (also known as PRSS6, SCCE) gene (Gene ID: 5650) in human. KLK7 is a member of the 15-member kallikrein subfamily of serine proteases. Human KLK7 displays 40 60% amino acid homology with other members of the family. Members of this family are involved in diverse physiological functions and serve as biomarkers for cancer. Increased expression of KLK7 is associated with unfavorable prognosis and progression of ovarian cancer. Less differentiated tumors of more advanced stage show higher level KLK7 expression. KLK7 has chymotrypsin-like activity and cleaves proteins with aromatic side chains in the P1 position. Its activity is inhibited by Zn2+ and Cu2+. KLK7 catalyzes the degradation of intercellular cohesive structures in the cornified layer of the skin in the continuous shedding of cells from the skin surface. It is also reported to play a role in the activation of precursors to inflammatory cytokines.

Specificity

Clone 73-1 specifically detects kallikrein 7 (KLK7) in human.

Immunogen

A full length recombinant human pro-Kallikrein 7.

Application

Detect KLK7 using this mouse monoclonal Anti-Kallikrein 7/KLK7 Antibody, clone 73-1, Cat. No. MABC967. It was used in ELISA and Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Kallikrein 7/KLK7 in human kidney tissue and human bone marrow.

ELISA Analysis: A representative lot detected Kallikrein 7/KLK7 in ELISA applications (Shan, S.J., et. al. (2006). Clin Chem. 52(10):1879-86; Kishi, T., et. al. (2004). Clin Chem. 50(4):709-16).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry in human skin tissue.

Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected Kallikrein 7/KLK7 in human skin tissue.

Target description

27.53 kDa calculated.

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Sodium bicarbonate buffer, pH 8.2 with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service