Skip to Content
Merck
All Photos(2)

Key Documents

MABC698

Sigma-Aldrich

Anti-PAWR, clone 3G9H7 Antibody

ascites fluid, clone 3G9H7, from mouse

Synonym(s):

PRKC apoptosis WT1 regulator protein, Prostate apoptosis response 4 protein, Par-4, PAWR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$562.00

$562.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$562.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$562.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3G9H7, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PAWR(5074)

General description

PRKC apoptosis WT1 regulator protein, also known as Prostate apoptosis response 4 protein (Par-4), and encoded by the gene name PAWR or PAR4, is a leucine zipper and death domain containing protein whose levels increase in neurons undergoing apoptosis as a result of trophic factor withdrawal or exposure to oxidative and metabolic insults. PAWR levels are reported to be increased in their lumbar spinal cord specimens further suggesting a role in neuronal degeneration. PAWR is localized to the cytoplasm in the absence of apoptosis signal and in normal cells and localized to the nucleus in most cancer cells. The tumor suppressor WT1 represses and activates transcription. The loss and/or imbalance of the dual transcriptional activity of WT1 may contribute to Wilms tumor. PAWR is a WT1 interacting protein that also functions as a transcriptional repressor.

Immunogen

Purified recombinant fragment of human PAWR expressed in E. Coli

Application

Detect PAWR using this Anti-PAWR, clone 3G9H7 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:200-1:1,000 dilution from a representative lot detected PAWR in human brain and breast carcinoma tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in Full-length Trx-PAWR recombinant protein and HeLa cell lysate.

Western Blotting Analysis: A 1:500-1:2,000 dilution of this antibody detected PAWR in Full-length Trx-PAWR recombinant protein and HeLa cell lysate.

Target description

~43 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid with up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service