Skip to Content
Merck
All Photos(2)

Key Documents

MABC682

Sigma-Aldrich

Anti-MATK Antibody, clone 9D7

ascites fluid, clone 9D7, from mouse

Synonym(s):

Megakaryocyte-associated tyrosine-protein kinase, CSK homologous kinase, CHK, Hematopoietic consensus tyrosine-lacking kinase, Protein kinase HYL, Tyrosine-protein kinase CTK

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$463.00

$463.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$463.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$463.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

9D7, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MATK(4145)

General description

Megakaryocyte-associated tyrosine-protein kinase (MATK), or alternatively CSK homologous kinase (CHK), Hematopoietic consensus tyrosine-lacking kinase, Protein kinase HYL, Tyrosine-protein kinase CTK, is an important kinase in hematopoietic signal transduction in blood cells as well as in other cells such as neurons and T cells. As its name would imply, MATK was first associated with megakaryocytes, the cells that give rise to platelets. MATK is able to phosphorylate and inactivate Src family kinases and may play a role in the regulation of proliferation of some cells such as T cells. In the nervous system MATK (called CHK in the older literature) plays a distinct role in the development of the CNS. MATK was found to be expressed in a developmental manner and expressed not only in developing neurons but also astrocytes and oligodendrocytes, but not in any of the associated tumors from these cell types, possibly indicating that it is the loss of expression of MATK activity that impacts tumorigenesis. EMD-Millipore’s Anti-MATK monoclonal antibody has been tested in western blot using K562 cell lysates and in flow cytometry using permeabilized K562 cells in culture.

Immunogen

Purified recombinant fragment of human MATK expressed in E. Coli.

Application

Detect Megakaryocyte-associated tyrosine-protein kinase using this mouse monoclonal antibody, Anti-MATK Antibody, clone 9D7 validated for use in western blotting & Flow Cytometry.
Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected MATK in K562 cells.

Optimal working dilutions must be determined by end user.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected MATK in K562 cell lysate.

Target description

~56 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified mouse monoclonal liquid in PBS containing up to 0.1% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
K562 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service