Skip to Content
Merck
All Photos(1)

Key Documents

MABC586

Sigma-Aldrich

Anti-Sulf-2 Antibody, clone 8G1

clone 8G1, from mouse

Synonym(s):

Extracellular sulfatase Sulf-2, hSulf-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$778.00

$778.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$778.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$778.00


Estimated to ship on24 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8G1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SULF2(55959)

General description

Sulf-2 is an endoplasmic reticulum enzyme that is involved in sulfur metabolism which is important in bone, cartilage, and skeletal development. Sulf-2 is also important in neurological situations including myelination and glial cell differentiation and GDNF activity. Sulf-2 activity is also found in the kidney and important in glomerular basement membrane development and glomerular function. Finally, Sulf-2 is involved in heparan sulfate metabolism. Sulf-2 removes sulfate moieties on heparan sulfate proteoglycans which in turn modulate the heparin binding growth factor signaling activity of the cell. Sulf-2 also impacts extracellular matrix interactions and it is important during development and growth, as well as Wnt signaling and VEGF production by endothelial cells during vascularization. In disease, Sulf-2 expression in cancer is upregulated and inhibition of Sulf-2 limits cancer progression and showed increased apoptosis within tumors, thus Sulf-2 activation is being examined as a potential anti-cancer therapy.

Immunogen

Recombinant protein corresponding to Human Sulf-2.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers
This Anti-Sulf-2 Antibody, clone 8G1 is validated for use in Western Blotting and Immunocytochemistry for the detection of Sulf-2.
Western Blot Analysis: 4 µg/mL of a representative lot of this antibody detected Sulf-2 in Sulf-2 transfected HEK293 cell lysates.

Immunocytochemistry Analysis: 4 µg/mL of a representative lot of this antibody detected Sulf-2 in Sulf-2 transfected HEK293 cells, but not in HEK293 cells transfected with an empty vector or Sulf-1.

Quality

Evaluated by Western Blotting in MCF-7 conditioned media.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Sulf-2 in 10 µg of MCF-7 conditioned media.

Target description

~125 & 75 kDa observed.
Sulf-2 is synthesized as a ~125 kDa pre-protein and is subsequently cleaved into ~75 kDa and ~50 kDa fragments, which are held together by disulfide bonds in the mature protein.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service