Skip to Content
Merck
All Photos(2)

Key Documents

MABC582

Sigma-Aldrich

Anti-hIAP-1/BIRC3 Antibody, clone 11D12.1

clone 11D12.1, from mouse

Synonym(s):

Baculoviral IAP repeat-containing protein 3, Apoptosis inhibitor 2, API2, C-IAP2, IAP homolog C, Inhibitor of apoptosis protein 1, IAP-1, hIAP-1, hIAP1, RING finger protein 49, TNFR2-TRAF-signaling complex protein 1, hIAP-1/BIRC3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$545.00

$545.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$545.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$545.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11D12.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BIRC3(330)

General description

Baculoviral IAP repeat-containing protein 3 (UniProt Q13489; also known as API2, Apoptosis inhibitor 2, Baculoviral IAP repeat-containing 3, c-IAP2, hIAP-1, hIAP1, IAP homolog C, IAP-1, Inhibitor of apoptosis protein 1, Mammalian IAP homolog C, RING finger protein 49, TNFR2-TRAF-signaling complex protein 1) is encoded by the BIRC3 (also known as API2, AIP1, IAP1, HAIP1, HIAP1, MALT2, MIHC, RNF49) gene (Gene ID 330) in human. hIAP-1 (c-IAP2) belongs to the inhibitor of apoptosis (IAP) family of proteins characterized by the presence of at least one baculoviral IAP repeat (BIR) domain. hIAP-1 (c-IAP2 contains three BIR (BIR1/2/3) domains, followed by an UBA (ubiquitin-associated), a CARD (caspase activation and recruitment domain), and a RING (really interesting new gene) domain. The BIR domains mediate hIAP-1 interaction with various binding partners, such as caspases, other IAP family members, and receptor-associated complexes. The RING domain and its associated E3 ligase mediates not only the ubiqutination of substrate proteins, such as caspase-3, caspase-7, SMAC/DIABLO, but also self-ubiquitylation. The UBA domain enables hIAP-1 to engage protein complexes containing poly-ubiquitylated protein(s).

Immunogen

GST-tagged recombinant protein corresponding to the BIR 1/2 of human hIAP-1/BIRC3.

Application

Detect BIRC3 using this mouse monoclonal Anti-hIAP-1/BIRC3 Antibody, clone 11D12.1, Cat. No. MABC582, validated for use in Western Blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected hIAP-1/BIRC3 in 10 µg of TF-1, Jurkat, and Molt-4 cell lysate.

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected hIAP-1/BIRC3 in 10 µg of K562 cell lysate.

Target description

~60 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service