Skip to Content
Merck
All Photos(2)

Key Documents

MABC550

Sigma-Aldrich

Anti-Pex3 Antibody, clone 14F3.2

Synonym(s):

Pex3|Peroxisomal biogenesis factor 3|Peroxin-3|Peroxisomal assembly protein PEX3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$743.00

$743.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

clone

monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

General description

Pex3 (also known as peroxin-3, peroxisomal assembly protein PEX3, peroxisomal biogenesis factor 3, transformation-related protein 18) is a multitransmembrane peroxisome protein that is encoded by the PEX3 gene in human (also known as PBD10A and TRG18). Pex19 is involved in maintaining peroxisome biosynthesis and integrity by acting as a docking factor for pex19 and, together with other peroxins (PEXs), is essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs), including Zellweger syndrome (ZWS), neonatal adrenoleukodystrophy (NALD), infantile Refsum disease (IRD), and classical rhizomelic chondrodysplasia punctata (RCDP), are lethal diseases characterized by multiple defects in peroxisome function, resulting in failure of importing one or more classes of peroxisomal matrix proteins into the organelle.

Immunogen

GST-tagged recombinant protein corresponding to human Pex3.

Application

Anti-Pex3 Antibody, clone 14F3.2 is an antibody against Pex3 for use in Western Blotting and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected Pex3 in human liver and human kidney tissue.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 ug/mL of this antibody detected Pex3 in 10 ug of HepG2 cell lysate.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service