Skip to Content
Merck
All Photos(1)

Key Documents

MABC521

Sigma-Aldrich

Anti-XBP1s Antibody, clone 143F (active form)

clone 143F, from mouse

Synonym(s):

X-box-binding protein 1, XBP-1, Tax-responsive element-binding protein 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$778.00

$778.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$778.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$778.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

143F, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... XBP1(7494)

General description

XBP-1, also known as X-box-binding protein 1 or Tax-responsive element-binding protein 5, and encoded by the gene XBP1/TREB5/XBP2, is a transcription factor essential for hepatocyte growth, the differentiation of plasma cells, immunoglobulin secretion, and the unfolded protein response (UPR). XBP-1 also acts during endoplasmic reticulum stress (ER) by activating unfolded protein response (UPR) target genes via direct binding to the UPR element (UPRE). XBP-1 binds DNA preferentially at CRE-like elements but also to some TPA response elements (TRE). XBP-1 is expressed in the nucleus. Defects in XBP-1 may be associated with Major affective disorder 7 (MAFD7) a major psychiatric disorder that is characterized by severe mood swings, with fluctuation between two abnormal mood states (manic or major depressive episode). Mania is accompanied by symptoms of euphoria, irritability, or excitation, whereas depression is associated with low mood and decreased motivation and energy.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of human XBP-1S, clone 143F (active form).

Application

Anti-XBP1s Antibody, clone 143F (active form) is an antibody against XBP1s for use in IHC & western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analyis: A representative lot from an independent laboratory detected XBP-1S, clone 143F (active form) in SK-MM-2, KARPAS 620, NCI-H929, and LP-1 cell lysates (Maestre, L., et al. (2009). Haematologica. 94(3):419-422.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected XBP-1S, clone 143F (active form) in human tonsil and lymphoplasmacytic cells (Maestre, L., et al. (2009). Haematologica. 94(3):419-422.).

Quality

Evaluated by Immunohistochemistry in human skin tissues.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected XBP-1S, clone 143F (active form) in human skin tissues.

Target description

29 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clara Lucía León-Annicchiarico et al.
The FEBS journal, 282(18), 3647-3658 (2015-07-15)
Altered metabolism is a hallmark of cancer that opens new therapeutic possibilities. 2-deoxyglucose (2-DG) is a non-metabolizable glucose analog tested in clinical trials and is frequently used in experimental settings to mimic glucose starvation. However, in the present study, conducted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service