Skip to Content
Merck
All Photos(3)

Key Documents

MABC34

Sigma-Aldrich

Anti-Beclin-1 Antibody, clone 9A1.1

clone 9A1.1, from mouse

Synonym(s):

Beclin-1, Coiled-coil myosin-like BCL2-interacting protein, Protein GT197

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9A1.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BECN1(8678)

General description

The Beclin-1 (BECN1) gene encodes a 60 kDa coiled-coil protein that interacts with the prototypic apoptosis inhibitor Bcl-2 (1). Beclin-1 promotes autophagy in autophagy-defective yeast with a targeted disruption of agp6/vps30, and in human MCF7 breast carcinoma cells. The autophagy-promoting activity of Beclin-1 in MCF7 cells is associated with inhibition of MCF7 cellular proliferation, in vitro clonigenicity, tumorigenesis, and has been found to ameliorate the neurodegenerative pathology in α-Synuclein models of Parkinson′s and lewy body diseases. Endogenous Beclin-1 protein expression is frequently low in human breast epithelial carcinoma cell lines and tissue, but is expressed ubiquitously at high levels in normal breast epithelia (2). It has also been shown that Beclin 1 contains a leucine-rich nuclear export signal motif raising the possibility that its autophagy and/or tumor suppressor function may require regulated, CRM1-dependent, nucleocytoplasmic transport (3).

Immunogen

GST-tagged recombinant protein corresponding to human Beclin-1.

Application

Anti-Beclin-1 Antibody, clone 9A1.1 detects level of Beclin-1 & has been published & validated for use in Western Blotting, IHC(P), Flow Cytometry.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Beclin-1 in human colorectal adenocarcinoma tissue.

Flow Cytometry Analysis: 1 µg from a representative lot detected Beclin-1 in HeLa cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in Raji cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Beclin-1 in 10 µg of Raji cell lysate.

Target description

~54 kDa kDa observed

Linkage

Replaces: MABN16

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cláudia Caldeira et al.
Frontiers in aging neuroscience, 9, 277-277 (2017-09-16)
Alzheimer's disease (AD) is characterized by a progressive cognitive decline and believed to be driven by the self-aggregation of amyloid-β (Aβ) peptide into oligomers and fibrils that accumulate as senile plaques. It is widely accepted that microglia-mediated inflammation is a

Articles

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service