Skip to Content
Merck
All Photos(2)

Documents

MABC272

Sigma-Aldrich

Anti-Annexin VI Antibody, clone 4B11.1

clone 4B11.1, from mouse

Synonym(s):

Annexin A6, 67 kDa calelectrin, Annexin VI, Annexin-6, Calphobindin-II, CPB-II, Chromobindin-20, Lipocortin VI, Protein III, p68, p70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B11.1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANXA6(309)

General description

The protein Annexin A6 or alternatively known as 67 kDa calelectrin, Annexin VI, Annexin-6, Calphobindin-II (CPB-II), Chromobindin-20, Lipocortin VI, Protein III, p68, or p70 and encoded by the human gene named ANXA6 or ANX6. Annexin VI is part of a large family of Annexin proteins and shares with the family the ability to bind negatively charged phospholipids in a calcium dependent manner. Localized in the plasma membrane annexins are important in many cellular functions as the provide scaffolds and play roles in vesicle transport, exocytosis, endocytosis and calcium transport and conduction. Recent research indicates that Annexin VI can interact with PKCalpha and EGFR in A431 cells to help regulate the phosphorylation of pY-EGFR and the associated activation of downstream signaling events associated with cancer. Annexin VI in normal tissue is expressed most strongly in myocardial tissue where it associates with sarcomeric α-actinin and is part of the sarcomere structure. Annexin VI also regulates terminal chondrocyte differentiation by modulating key signaling pathway activities.

Immunogen

GST-tagged recombinant protein corresponding to human Annexin VI.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Annexin VI Antibody, clone 4B11.1 is validated for use in western blotting & ICC for the detection of Annexin VI.
Western Blotting Analysis: 1 µg/mL from a representative lot detected Annexin VI in 10 µg of C2C12 cell lysate.

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Annexin VI in HeLa and A431 cells.

Alexa Fluor is a registered trademark of Life Technologies.

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Annexin VI in 10 µg of SH-SY5Y cell lysate.

Target description

~75 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service