Skip to Content
Merck
All Photos(1)

Key Documents

MABC1798

Sigma-Aldrich

Anti-Claudin-6 Antibody, clone 2

Synonym(s):

Skullin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$233.00
100 μL
$590.00

$233.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$233.00
100 μL
$590.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$233.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2, monoclonal

species reactivity

human

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

Gene Information

human ... CLDN6(9074)

Specificity

Clone 2 is a mouse monoclonal antibody that detects Claudin-6. It targets an epitope within the extracellular domain.

Immunogen

Extracellular peptides specific for human Claudin-6.

Application

Quality Control Testing

Evaluated by Western Blotting in NTERA-2 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Claudin-6 in NTERA-2 cell lysate.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected Claudin-6 in Immunocytochemistry applications (Kong, F-E., et al. (2021). Sci Transl Med. 13(579):eabb6282).

Inhibition Assay: A representative lot inhibited tumor growth of HepG2 xenograft tumors in a murine model. (Kong, F-E., et al. (2021). Sci Transl Med.;13(579):eabb6282).

Immunohistochemistry Applications: A representative lot detected Claudin-6 in Immunohistochemistry applications (Kong, F-E., et al. (2021). Sci Transl Med. 13(579):eabb6282).

Western Blotting Analysis: A representative lot detected Claudin-6 in Western Blotting applications (Kong, F-E., et al. (2021). Sci Transl Med. 13(579):eabb6282).

Flow Cytometry Analysis: A representative lot detected Claudin-6 in Flow Cytometry applications (Kong, F-E., et al. (2021). Sci Transl Med. 13(579):eabb6282).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Claudin-6 (UniProt: P56747; also known as Skullin) is encoded by the CLDN6 (also known as UNQ757/PRO1488) gene (Gene ID: 9074) in human. Claudin-6 is a multi-pass membrane protein that plays a major role in tight junction-specific obliteration of the intercellular space. Claudin-6 has two extracellular domains (aa 29-81 and 138-160), four transmembrane domains (aa 8-28, 82-102, 117-137, and 161-181), and three cytoplasmic domains (aa 1-7; 103-116, and 182-220). It acts as a receptor for hepatitis C virus (HCV) entry into hepatic cells. Claudin-6 is expressed in the liver, in peripheral blood mononuclear cells, and hepatocarcinoma cell lines. It is also expressed specifically in undifferentiated stem cells and can serve as a marker to purify stem-like cells from a mixed cell population. It is shown to play an important role during mouse embryonic epithelium formation and the development of endodermal tissues, and its level of expression is correlated with dedifferentiation and malignant proliferation of epithelial cells. (Ref.: Kong, F-E. et al. (2021). Sci. Transl. Med. 13, eabb6282; Wang, L., et al. (2012). Cell Res. 22; 1082-1085; Zheng, A., et al. (2007). J. Virol. 81(22); 12465-12471).

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service