Skip to Content
Merck
All Photos(2)

Key Documents

MABC1558

Sigma-Aldrich

Anti-SPMD3 (nSMase2) Antibody, clone 14G5.1

clone 14G5.1, from mouse

Synonym(s):

Sphingomyelin phosphodiesterase 3, EC: 3.1.4.12, Neutral sphingomyelinase 2, nSMase-2, nSMase2, Neutral sphingomyelinase II

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$252.00

$252.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$252.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

14G5.1, monoclonal

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SMPD3(55512)

General description

Sphingomyelin phosphodiesterase 3 (UniProt: Q9NY59; also known as EC: 3.1.4.12, Neutral sphingomyelinase 2, nSMase-2, nSMase2, Neutral sphingomyelinase II) is encoded by the SMPD3 gene (Gene ID: 55512) in human. SMases are classified based on their pH optimum into acid, neutral, and alkaline subtypes. Four different neutral SMases have been described in mammals and nSMase-2 is shown to be the most predominant in cellular systems. nSMase-2 is a phosphoprotein whose activity is regulated by calcineurin (PP2B) and it is shown to be exclusively phosphorylated on serine residues. It is predominantly expressed in the brain and its levels are shown to be up-regulated during G0/G1 phases. Two isoforms of nSMase-2 are reported that are produced by alternative splicing. nSMase-2 catalyzes the generation of bioactive lipid ceramide through the hydrolysis of the membrane lipid sphingomyelin and specifically hydrolyzes the phosphocholine-headgroup from sphingomyelin. It does not exhibit phospholipase C-type activity against phosphatidylcholine, lysophosphatidylcholine, platelet activating factor or lyso-platelet activating factor. For its activity a neutral pH and divalent cations (Mg2+ or Mn2+) are required and its activity is stimulated by the presence of phosphatidylserine (PS) and unsaturated fatty acids. NSMase-2 undergoes palmitoylation and palmitoylation-deficient protein is target for lysosomal degradation. (Ref.: Filosto, S., et al (2010). J. Biol. Chem. 285 (14); 10213-10222; Shamseddine, A.A., et al (2013). Adv. Enz. Regul. 57: 24-41).

Specificity

Clone 14G5.1 detects neutral sphingomyelinase 2 in human and murine brain tissue. It targets an epitope within 101 amino acids from the C-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 101 amino acids from the C-terminal region of human Neutral sphingomyelinase 2.

Application

Anti-SPMD3 (nSMase2), clone 14G5.1, Cat. No. MABC1558, is a mouse monoclonal antibody that detects Sphingomyelin phosphodiesterase 3 (sSMase) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SPMD3 (nSMase2) in human brain tissue lysate.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected SPMD3 (nSMase2) in human stomach and human fallopian tube tissue sections.

Quality

Evaluated by Western Blotting in mouse cerebellum tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SPMD3 (nSMase2) in mouse cerebellum tissue lysate.

Target description

~70 kDa observed; 71.08 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Mouse monoclonal antibody in tissue culture supernatant without azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service