Skip to Content
Merck
All Photos(1)

Key Documents

MABC1182

Sigma-Aldrich

Anti-Serpina3n Antibody, clone 13H5

clone 13H5, from hamster(Armenian)

Synonym(s):

Serine protease inhibitor A3N

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$556.00

$556.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$556.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$556.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

hamster (Armenian)

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13H5, monoclonal

species reactivity

mouse

should not react with

human

technique(s)

immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Serine protease inhibitor A3N (UniProt: Q91WP6; also known as Serpin A3N) is encoded by the Serpina3n (also known as Spi2) gene (Gene ID: 20716) in murine species. SerpinA3N is a secreted protein of the serpin family. Serpins bind and inhibit specific serine proteases and several serine proteases are reported to be substrates for SerpinA3N in vitro. It is synthesized with a signal peptide of 20 amino acids that is cleaved to generate the mature protein. SerpinA3N is expressed at high levels in brain, heart, liver, lung, spleen, testis, and thymus. However, its expression is low in kidney, skeletal muscle, and bone marrow. Serpin A3N expression can also be induced by bacterial lipopolysaccharides. The reactive center loop (RCL) of as SerpinA3N extends out from the body of the protein and directs binding to the target protease. The protease cleaves the serpin at the reactive site within the RCL, establishing a covalent linkage between the serpin reactive site and the protease. The resulting inactive serpin-protease complex is highly stable. SerpinA3N is shown to be up-regulated in the dorsal root ganglia after nerve injury and mice lacking SerpinA3N are reported to develop more neuropathic mechanical allodynia than wild-type (WT) mice. SerpinA3N is also shown to attenuate activated T cell-mediated neuronal death and reduce the severity of disease in experimental autoimmune encephalomyelitis (EAE). (Ref.: Vicuna, L et al. (2015) Nat. Med. 21(5):518-23).

Specificity

Clone 13H5 specifically targets murine Serpina3n. It is predicted to react with both pro- and mature forms of Serpina3n. This clone targets a sequence within 24 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal region of murine Serpina3n.

Application

Detect Serpina3n using this armenian hamster monoclonal Anti-Serpina3n Antibody, clone 13H5, Cat. No. MABC1182. It has been tested for use in Immunofluorescence and Western Blotting.
Immunofluorescence Analysis: A representative lot detected increased Serpin A3N/Serpina3n immunoreactivity in late metastasis stage lung tissue by fluorescent immunohistochemistry staining of paraformaldehyde-fixed frozen lung sections in a study employing the murine MMTV-PyMT breast-to-lung metastasis model (Sevenich, L., et al. (2014). Nat. Cell Biol. 16(9):876-888).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Serpina3n in 10 µg of mouse brain tissue lysate.

Target description

~46 kDa observed; 46.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified armenian hamster monoclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehwish Saba Aslam et al.
BioMed research international, 2022, 1557010-1557010 (2022-06-10)
Ischemic stroke is a devastating CNS insult with few clinical cures. Poor understanding of underlying mechanistic network is the primary limitation to develop novel curative therapies. Extracellular accumulation of granzyme B subsequent ischemia promotes neurodegeneration. Inhibition of granzyme B can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service