Skip to Content
Merck
All Photos(1)

Key Documents

MABC1147

Sigma-Aldrich

Anti-CIP2A Antibody, clone 6G8-G1

clone 6G8-G1, from mouse

Synonym(s):

Protein CIP2A, Cancerous inhibitor of PP2A, p90 autoantigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$506.00

$506.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$506.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$506.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G8-G1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Protein CIP2A (UniProt: Q8TCG1; also known as CIP2A, Cancerous inhibitor of PP2A, p90 autoantigen) is encoded by the KIAA1524 (also known as CIP2A) gene (Gene ID: 57650) in human. CIP2A is a single pass membrane protein that exhibits dynamic changes in localization, including the cytoplasm and centrosome, depending on the stage of the cell cycle. It is expressed at low levels in most tissues, however, it is overexpressed in several cancers, including head-and-neck squamous cell carcinomas and liver cancer cells. Levels of CIP2A are correlated with the proliferation of human cancer cells and with cancer progression. It acts as an oncoprotein and inhibits protein phosphatase 2A activity and thereby stabilizes c-Myc in human malignancies. It promotes anchorage-independent cell growth and tumor formation. Higher activity of c-Myc rapidly induces the activation of cyclin E-cdk2 kinase activity. Concurrently, the release of p27Kip1 from Cdk2 complexes enhances cell proliferation. In CIP2A depleted cells, centrosome separation and the mitotic spindle dynamics were impaired, which leads to the activation of spindle assembly checkpoint signaling and, ultimately, delayed cell division time. CIP2A is also reported to interact with NIMA (never in mitosis gene A)-related kinase 2 (NEK2) during G2/M phase and enhances NEK2 kinase activity that facilitates centrosome separation in a PP1- and PP2A-independent manner. (Ref.: Jeong, AL et al. (2014). J. Biol. Chem. 289(1): 28-40).

Specificity

Clone 6G8-G1 specifically detects CIP2A in HEK293 cells overexpressing CIP2A.

Immunogen

Epitope: N-terminus
His-tagged recombinant fragment from the N-terminal region of human CIP2A.

Application

Detect CIP2A using this mouse monoclonal Anti-CIP2A Antibody, clone 6G8-G1, Cat. No. MABC1147, tested in Western Blotting.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in HEK293 cells overexpressing CIP2A.

Western Blotting Analysis: 1 µg/mL of this antibody detected CIP2A in 10 µg of lysate from HEK293 cells overexpressing CIP2A.


Target description

~102 kDa observed; 102.18 calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service