Skip to Content
Merck
All Photos(2)

Key Documents

MABC1127

Sigma-Aldrich

Anti-FGFR-4 Antibody, clone 29.2

culture supernatant, clone 29.2, from rabbit

Synonym(s):

Fibroblast growth factor receptor 4, EC: 2.7.10.1, CD334

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$214.00
100 μL
$566.00

$214.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$214.00
100 μL
$566.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

culture supernatant

antibody product type

primary antibodies

clone

29.2, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... FGFR4(2264)

General description

Fibroblast growth factor receptor 4 (UniProt: P22455; also known as EC: 2.7.10.1, FGFR-4, CD334) is encoded by the FGRF4 (also known as JTK2, TKF) gene (Gene ID: 2264) in human. FGFR-4 is a highly conserved, single-pass type I membrane protein with tyrosine kinase activity that serves as a cell surface receptor for fibroblast growth factors and regulates cell proliferation differentiation, and migration. FGFR-4 is expressed in gastrointestinal epithelial cells, pancreas, and gastric and pancreatic cancer cell lines. In the absence of a ligand, it is present as a monomer in an inactive conformation. Upon ligand binding it homodimerizes into an active conformation with autophosphorylation of tyrosine residues. FGFR-4 is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off in the mature form. It contains an extracellular domain (aa 22-369), a transmembrane domain (aa 370-390), and a cytoplasmic domain (aa 391-802). FGFR-4 undergoes N-glycosylation, and when not fully glycosylated it can undergo proteasomal degradation. FGFR-4 signaling is down-regulated by receptor internalization and degradation. MMP14 is reported to promote internalization and degradation of FGFR-4. Mutations that lead to constitutive kinase activation or impair normal FGFR-4 inactivation lead to aberrant signaling leading to tumor growth. (Ref.: Turkington, RC et al. (2014). Cell Death Dis. 5; e1046).

Specificity

This rabbit monoclonal antibody detects human fibroblast growth factor receptor 4 (FGFR-4). It targets an epitope with in the extracellular domain.

Immunogen

Epitope: extracellular domain
His-tagged recombinant extracellular domain of human fibroblast growth factor receptor 4 (FGFR-4).

Application

Anti-FGFR-4, clone 29.2, Cat. No. MABC1127, is a rabbit monoclonal antibody that detects Fibroblast growth factor receptor 4 and has been tested for use in Flow Cytometry.
Flow Cytometry Analysis: A 1:4 dilution from a representative lot detected FGFR-4 in one million LNCaP cells.
Research Category
Signaling

Quality

Evaluated by Flow Cytometry in DU-145 cells.

Flow Cytometry Analysis: A 1:4 dilution of this antibody detected FGFR-4 in one million DU-145 cells.

Target description

87.95 kDa calculated.

Physical form

Rabbit monoclonal antibody in supernatant without preservatives.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service