Skip to Content
Merck
All Photos(1)

Key Documents

MABC002H

Sigma-Aldrich

Mouse IgG1 Negative Control, clone Ci4, PE conjugate

Mouse IgG1 Negative Control Monoclonal Antibody validated for use in Flow Cytometry.

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$485.00

$485.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$485.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$485.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

clone

Ci4, monoclonal

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Specificity

This antibody reacts with chicken immunoglobulin. No reaction with human cell surface or plasma components has been observed. However some binding to Fc receptors may occur.

Immunogen

Chicken immunoglobulin

Application

Mouse IgG1 Negative Control Monoclonal Antibody validated for use in Flow Cytometry.
Phycoerythrin conjugated mouse IgG1 is a monoclonal antibody intended for use as a negative control in flow cytometry. Its use enables an estimation of non-specific binding of phycoerythrin conjugated mouse monoclonal antibodies to cell surface components in peripheral blood.

SUGGESTED USAGE

Flow cytometry - use 10 μl direct from the vial per 100 μl of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) or bone marrow cells in 100 μl buffer.
Research Category
Secondary & Control Antibodies

Epitope Tags & General Use
Research Sub Category
Isotype Control Antibodies

Physical form

Purified immunoglobulin. R-Phycoerythrin* conjugatedready to use. The antibody is supplied in 1.0ml phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Storage and Stability

Store at 2 to 8°C. DO NOT FREEZE. Protect from light, for up to 6 months.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Fritah et al.
Cancers, 12(9) (2020-09-16)
Resistance to chemotherapy by temozolomide (TMZ) is a major cause of glioblastoma (GBM) recurrence. So far, attempts to characterize factors that contribute to TMZ sensitivity have largely focused on protein-coding genes, and failed to provide effective therapeutic targets. Long noncoding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service