Skip to Content
Merck
All Photos(1)

Documents

MAB8515

Sigma-Aldrich

Anti-Human Metapneumovirus 75.1 Antibody, clone 2D7

clone 2D7, Chemicon®, from mouse

Synonym(s):

Metapneumovirus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

2D7, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG2b

shipped in

wet ice

Specificity

Human Metapneumoviruses. Reacts against both A and B antigenic groups and subgroups 1 and 2

Immunogen

Epitope: localized to the M1 protein human metapneumovirus
MPV CAN98-75

Application

ELISA

Immunofluorescence:



It is difficult to grow the metapneumovirus in standard cell lines. LLC-MK2 can be used for culture; acetone fixation is recommended. See Ebihara, 2005.



Optimal dilutions must be determined by the end user
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Human Metapneumovirus 75.1 Antibody, clone 2D7 is validated for use in ELISA, IF for the detection of Human Metapneumovirus 75.1.

Physical form

Format: Purified
Purified immunoglobulin. 0.02 M Phosphate Buffered Saline, 0.25 M Sodium Chloride, 0.1% Sodium Azide, pH = 7.6

Storage and Stability

Store at 2° to 8°C for up to 1 year from date of receipt

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service