Skip to Content
Merck
All Photos(1)

Key Documents

MAB706

Sigma-Aldrich

Anti-E. coli Antibody, 02 Antigen, Non-Laboratory Stains, clone 1002/224.15

clone 1002/224.15, Chemicon®, from mouse

Synonym(s):

Escherichia coli

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$407.00

$407.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$407.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$407.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1002/224.15, monoclonal

species reactivity

E. coli

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG3

shipped in

wet ice

Specificity

E. coli expressing the polysaccharide antigen 02.

Immunogen

Epitope: 02 Antigen, Non-Laboratory Stains

Application

Detect the E. coli protein using this Anti-E. coli, 02 Antigen, Non-Laboratory Stains, clone 1002/224.15 validated for use in ELISA & IF.
ELISA

IFA

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Bacterial

Physical form

Format: Purified
Purified immunoglobulin in 0.02M, 0.25M NaCl, pH7.6, with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Uemura et al.
International journal of laboratory hematology, 31(1), 36-47 (2008-02-05)
We diagnosed an 86-year-old woman with chronic neutrophilic leukemia (CNL) because she showed sustained leukocytosis dominated by mature neutrophils, hepatosplenomegaly, high neutrophilic alkaline phosphatase score, absence of the Ph chromosome, low serum level of granulocyte colony-stimulating factor (G-CSF) and granulocyte-macrophage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service