Skip to Content
Merck
All Photos(1)

Key Documents

MAB5674

Sigma-Aldrich

Anti-Plectin Antibody, Clone 10F6

clone 10F6, Chemicon®, from mouse

Synonym(s):

Plackin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$785.00

$785.00


Estimated to ship on31 May 2025



Select a Size

Change View
100 μG
$785.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$785.00


Estimated to ship on31 May 2025


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10F6, monoclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

The product of this gene belongs to the plakin or cytolinker family of genes. The encoded protein is involved in cytoskeleton-membrane attachment in epithelial cells, muscle, and other tissues. It also may regulate changes of lymphocyte cytoarchitecture during polarization and extravasation. This gene has alternative splicing of different 5′ exons to the first constant exon, which generates transcript variants that encode distinct isoforms

Specificity

Cat. # MAB5674 recognizes the cytoplasmic domain of Plectin.
Reactivity with other species has not been tested.

Immunogen

Gel-purified C5 Cell plectin

Application

Detect Plectin using this Anti-Plectin Antibody, Clone 10F6 validated for use in WB, IH.
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Neuronal & Glial Markers

Quality

Routinely tested on rat brain tissue lysate

Target description

300-500 kDa

Physical form

Format: Purified
Purified in PBS with 0.05% NaN3

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Ambrosini et al.
Glia, 41(3), 290-300 (2003-01-16)
Macrophage inflammatory protein-3alpha/CCL20 is a recently identified chemokine that binds to CCR6 and acts as a chemoattractant for memory/differentiated T-cells, B-cells, and immature dendritic cells. We have previously reported that CCL20 and CCR6 mRNAs are expressed in the CNS of
Sarah D Ackerman et al.
The Journal of experimental medicine, 215(3), 941-961 (2018-01-26)
Myelin is a multilamellar sheath generated by specialized glia called Schwann cells (SCs) in the peripheral nervous system (PNS), which serves to protect and insulate axons for rapid neuronal signaling. In zebrafish and rodent models, we identify GPR56/ADGRG1 as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service