MAB5628
Anti-Glial Fibrillary Acidic Protein Antibody
CHEMICON®, mouse monoclonal,
Synonym(s):
GFAP
Select a Size
$738.00
Estimated to ship on28 April 2025
Select a Size
About This Item
$738.00
Estimated to ship on28 April 2025
Recommended Products
Product Name
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from mouse
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
monoclonal
species reactivity
rat, human
manufacturer/tradename
Chemicon®
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... GFAP(2670)
Application
Optimal working dilutions must be determined by end user.
Linkage
Physical form
Analysis Note
Positive: Neural tissue or astrocyte culture
Negative: Non-neural tissue or neuronal culture
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service