Skip to Content
Merck
All Photos(1)

Documents

MAB5580

Sigma-Aldrich

Anti-Arrestin Antibody, visual

ascites fluid, Chemicon®

Synonym(s):

S-antigen, rod arrestin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

rat, human, pig, bovine, goat, sheep

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SAG(6295)

General description

Visual arrestin is a 48 kDa protein expressed exclusively in retinal photoreceptor cells where it regulates light-dependent signal transduction through G protein-coupled receptor (rhodopsin) activation. Visual arrestin is the major pathogenetic autoantigen in human autoimmume imflammatory eye disease.

Specificity

Visual arrestin. The antibody has not been tested for reactivity to arrestin in other tissues.

Immunogen

Porcine visual arrestin.

Application

Detect Arrestin using this Anti-Arrestin Antibody, visual validated for use in IH, IP & WB.
Research Category
Neuroscience
Research Sub Category
Sensory & PNS
Western blot: 48kDa

Immunohistochemistry

Immunopurification of arrestin

Optimal working dilutions must be determined by end user.

Physical form

Ascites fluid. Liquid.

Storage and Stability

Maintain -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Chang et al.
Frontiers in pharmacology, 14, 1240829-1240829 (2023-12-21)
Introduction: Pulmonary fibrosis (PF) is a fatal chronic lung disease that causes structural damage and decreased lung function and has a poor prognosis. Currently, there is no medicine that can truly cure PF. Vitamin E (VE) is a group of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service