Skip to Content
Merck
All Photos(1)

Key Documents

MAB5572

Sigma-Aldrich

Anti-NMDAR2A Antibody

ascites fluid, clone 2H9.24A6, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$632.00

$632.00


Estimated to ship on24 April 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB04901

Request a Bulk Order

Select a Size

Change View
100 μL
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Estimated to ship on24 April 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB04901

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

2H9.24A6, monoclonal

species reactivity

frog, rat, human, rabbit, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIN2A(2903)

Specificity

NMDAR2A, N-terminal. By Western blot the antibody reacts with bands at ~165 and ~100 kDa on rat brain lysate.

Immunogen

Recombinant protein from the N-terminal of rat NMDAR2A .

Application

Anti-NMDAR2A Antibody detects level of NMDAR2A & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:100-1:500 on rat brain lysate.

Optimal working dilutions must be determined by end user.

Physical form

Liquid

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Rat forebrain or cerebellum.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pilocapine alters NMDA receptor expression and function in hippocampal neurons: NADPH oxidase and ERK12 mechanisms.
Di Maio R, Mastroberardino PG, Hu X, Montero L, Greenamyre JT
Neurobiology of Disease null
D J Laurie et al.
Brain research. Molecular brain research, 51(1-2), 23-32 (1998-01-14)
Mouse monoclonal antibodies were raised against bacterially expressed protein sequences of the NR2A, NR2B, NR2C and NR2D subunits of the rat NMDA receptor. From immunoblots of rat brain proteins, the apparent molecular weights of these subunits were 165, 170, 135
Verónica Guirao et al.
Journal of neurochemistry, 143(3), 359-374 (2017-09-08)
The statin atorvastatin (ATV) given as a post-treatment has been reported beneficial in stroke, although the mechanisms involved are not well understood so far. Here, we investigated in vitro the effect of post-treatment with ATV and its main bioactive metabolite ortho-hydroxy
Dick Terwel et al.
Journal of neurochemistry, 118(1), 105-112 (2011-04-27)
Wilson's disease (WD) is caused by mutations in the copper transporting ATPase 7B (Atp7b). Patients present with liver pathology or behavioural disturbances. Studies on rodent models for WD so far mainly focussed on liver, not brain. The effect of knockout

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service