Skip to Content
Merck
All Photos(1)

Key Documents

MAB5566

Sigma-Aldrich

Anti-Tubulin α Antibody, detyrosinated

ascites fluid, Chemicon®

Synonym(s):

Anti-Anti-B-ALPHA-1, Anti-Anti-LIS3, Anti-Anti-TUBA3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$716.00


Estimated to ship on02 June 2025


biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

Specificity

Alpha tubulin, detyrosinated.

Immunogen

Epitope: detyrosinated
Synthetic peptide GEEEGE.

Application

(add note)
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers
This Anti-Tubulin Antibody, α, detyrosinated is validated for use in WB for the detection of Tubulin.

Physical form

Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-intensity pulsed ultrasound accelerates tooth movement via activation of the BMP-2 signaling pathway.
Xue, H; Zheng, J; Cui, Z; Bai, X; Li, G; Zhang, C; He, S; Li, W; Lajud, SA; Duan, Y; Zhou, H
Testing null
Yu-Ming Lu et al.
eLife, 13 (2024-07-01)
Tubulin posttranslational modifications (PTMs) modulate the dynamic properties of microtubules and their interactions with other proteins. However, the effects of tubulin PTMs were often revealed indirectly through the deletion of modifying enzymes or the overexpression of tubulin mutants. In this
Fan Zhang et al.
Translational neurodegeneration, 4, 9-9 (2015-06-02)
In Alzheimer disease (AD), hyperphosphorylation of tau proteins results in microtubule destabilization and cytoskeletal abnormalities. Our prior ultra-morphometric studies documented a clear reduction in microtubules in pyramidal neurons in AD compared to controls, however, this reduction did not coincide with
Leticia Peris et al.
Brain : a journal of neurology (2022-02-12)
Microtubules play fundamental roles in the maintenance of neuronal processes and in synaptic function and plasticity. While dynamic microtubules are mainly composed of tyrosinated tubulin, long-lived microtubules contain detyrosinated tubulin, suggesting that the tubulin tyrosination/detyrosination cycle is a key player
Panseon Ko et al.
International journal of molecular sciences, 22(11) (2021-07-03)
During aggressive cancer progression, cancer cells adapt to unique microenvironments by withstanding various cellular stresses, including endoplasmic reticulum (ER) stress. However, the mechanism whereby cancer cells overcome the ER stress to survive remains to be elucidated. Herein, we demonstrated that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service