Skip to Content
Merck
All Photos(1)

Key Documents

MAB5292

Sigma-Aldrich

Anti-Dopamine D1B Receptor Antibody, clone SG4-D1B

clone SG4-D1B, Chemicon®, from mouse

Synonym(s):

Dopamine D5 Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Product MAB5292 is not currently sold in your country. Contact Technical Service

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SG4-D1B, monoclonal

species reactivity

mouse, rat

should not react with

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DRD5(1816)

Specificity

Dopamine Receptor D1b (D5).

Immunogen

Recombinant rat D1b Receptor, carboxy terminus.

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-Dopamine D1B Receptor Antibody, clone SG4-D1B is validated for use in IH, WB for the detection of Dopamine D1B Receptor.
Western blot: 1:250-1:500 on mouse brain lysate (unboiled sample)

Immunohistochemistry

Optimal working dilutions must be determined by the end user.

SUGGESTED WESTERN BLOT PROTOCOL

Solutions

10X Transfer buffer 1 liter

Tris 115.3 gm

Glycine 24.2 gm

[SDS 4 gm (may be omitted)]

For a 1X working solution: 1 liter

mix 700 mL water

200 mL

MeOH100 mL

10X Transfer

TBSTstock 1 liter1M

Tris pH 7.5 10 mL

5 M NaCl 30 mL

10% Tween-20 5 Ml

1. Run SDS-PAGE gel as desired.

2. Soak PVDF membrane (ImmobilonP from Millipore) in 100% methanol for 1-2 minutes to wet the membrane.

3. Soak membrane in transfer buffer (see below) until ready to set up blot (>15 min) transfer.

4. Assemble gel-transfer sandwich with the blot toward the positive electrode.

5. Transfer protein from gel to membrane at 100 volts for 1-2 hours for small molecular weight proteins or 65 mA for 16 hours (overnight) for complete transfer of higher molecular weight proteins.

6. Stain the transferred bands with Chemicon BLOT-FastStain (Catalog Number 2076).

7. Destain with deionized water.

8. Block with 5% non-fat milk (Marvel or Carnation) in water, overnight at 4°C. The non-fat milk should be dissolved freshly, centrifuged 10,000 rpm for 10 min, and filtered through glass filter (Gelman Acrodisc).

9. Incubation with first antibody overnight with rocking at 4°C in blocking solution. Optimal working dilutions and incubation time will need to be determined by the end user.

10. Wash at least 3 x 5 min. with TBST. From this stage, azide should be omitted.

11. Incubation with the secondary antibody (HRP-conjugated goat anti-mouse antibody, for example Chemicon Catalog Number AP124P, diluted appropriately) 1 h at room temperature.

12. Wash at least 3 x 5 min. with TBST.

13. Perform ECL with commercial kit (femtoLUCENT, Chemicon Catalog Number 2078).

Linkage

Replaces: AB1791P

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02 M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Usman M Ashraf et al.
Biomedicines, 11(1) (2023-01-22)
The genetic and molecular basis of developing high blood pressure and renal disease are not well known. Resp18mutant Dahl salt-sensitive (SS-Resp18mutant) rats fed a 2% NaCl diet for six weeks have high blood pressure, increased renal fibrosis, and decreased mean
Carlo Cavallotti et al.
Neurochemical research, 29(8), 1499-1504 (2004-07-21)
Dopamine receptors (Dar) were studied as a component of the nervous dopaminergic system in the human dura mater. Dar were stained in several dural zones (vascular, perivascular, intervascular) in different regions (basal, calvarial, tentorial, occipital, frontal, parietal, temporal) of the
Vito S Hernández et al.
Neuropharmacology, 82, 88-100 (2013-12-03)
L-Dopa is the major symptomatic therapy for Parkinson's disease, which commonly occurs in elderly patients. However, the effects of chronic use on mood and cognition in old subjects remain elusive. In order to compare the effects of a chronic pulsatile
Balaji Krishnan et al.
PloS one, 6(9), e25639-e25639 (2011-10-08)
Cocaine-cue associations induce synaptic plasticity with long lasting molecular and cellular changes in the amygdala, a site crucial for cue-associated memory mechanisms. The underlying neuroadaptations can include marked alterations in signaling via dopamine (DA) receptors (DRs) and metabotropic glutamate (Glu)
William Galbavy et al.
Molecular pain, 9, 60-60 (2013-11-29)
Dopaminergic fibers originating from area A11 of the hypothalamus project to different levels of the spinal cord and represent the major source of dopamine. In addition, tyrosine hydroxylase, the rate-limiting enzyme for the synthesis of catecholamines, is expressed in 8-10%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service