Skip to Content
Merck
All Photos(1)

Key Documents

MAB4407

Sigma-Aldrich

Anti-HESCA-1 Antibody, clone 051007-4A5

clone 051007-4A5, Chemicon®, from mouse

Synonym(s):

Human Embryonic Stem Cell Antigen-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$807.00

$807.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$807.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$807.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

051007-4A5, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

input

sample type: human embryonic stem cell(s)
sample type induced pluripotent stem cell(s)

isotype

IgMκ

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

The evolution of our understanding of pluripotent and adult stem cell biology requires specific research tools to permit the characterization, isolation, and expansion of stem cells. HESCA-1, developed in collaboration with Abeome Corporation, is the first in a series of novel, human embyronic stem cell antibodies produced to provide scientists with those needed tools. This novel monoclonal antibody recognizes a newly discovered 200 kDa cell surface protein that has been shown to be expressed in pluripotent human embryonic stem cell (hESC) populations.

Specificity

Reactivity to other species has not been determined.
This antibody is positive by immunofluorescence against H1, BG-01, BG01v, and BF02 human embryonic stem (hES) cell lines; however, it is unreactive for differentiated stem cells in teratomas and common feeder lines such as MEF, D-551, and Hs27. Reactivity with other hES lines has not yet been determined.

Immunogen

BG01v hES cell line

Application

Detect HESCA-1 (Human Embryonic Stem Cell Antigen-1) using this Anti-HESCA-1 Antibody, clone 051007-4A5 validated for use in IC, IH, IF, FC.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Target description

200 kDa

Physical form

Format: Purified
Liquid in 0.02M PBS, 0.25 NaCl, pH 7.6 with 0.1% Sodium Azide

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 1 year from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service