Skip to Content
Merck
All Photos(1)

Key Documents

MAB396

Sigma-Aldrich

Anti-Glutamate Receptor 2 & 4 Antibody, clone 3A11

clone 3A11, Chemicon®, from mouse

Synonym(s):

GluR2/4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$683.00

$683.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$683.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A11, monoclonal

species reactivity

monkey, human, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIA2(2891)
rat ... Gria2(29627)
rhesus monkey ... Gria2(574344)

Specificity

Glutamate Receptor 2 (GluR2) by Western blot and RIA. When using MAB396 for immunohistochemistry or immunoprecipitation it recognizes both GluR2 and GluR4.

Immunogen

Recombinant fusion protein TrpE-GluR2

Application

Detect Glutamate Receptor 2 & 4 using this Anti-Glutamate Receptor 2 & 4 Antibody, clone 3A11 validated for use in ELISA, IC, IH, IP, RIA & WB.
Immunohistochemistry on 4% paraformaldehyde fixed

tissue: 1:500-1,000

Immunocytochemistry

ELISA

RIA

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS, no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Chen et al.
Journal of neuroscience research, 81(4), 522-529 (2005-06-11)
Lipid rafts are specialized plasma membrane microdomains enriched in cholesterol and sphingolipids that serve as major assembly and sorting platforms for signal transduction complexes. Constitutively expressed heat shock proteins Hsp90, Hsc70, Hsp60, and Hsp40 and a range of neurotransmitter receptors
Dannis G van Vuurden et al.
PloS one, 4(6), e5953-e5953 (2009-06-19)
Glioblastoma multiforme (GBM) cells secrete large amounts of glutamate that can trigger AMPA-type glutamate receptors (AMPARs). This commonly results in Na(+) and Ca(2+)-permeability and thereby in excitotoxic cell death of the surrounding neurons. Here we investigated how the GBM cells
Shoko Shimizu et al.
Glia, 62(5), 709-724 (2014-02-01)
Recent studies have shown changes in myelin genes and alterations in white matter structure in a wide range of psychiatric disorders. Here we report that DBZ, a central nervous system (CNS)-specific member of the DISC1 interactome, positively regulates the oligodendrocyte
Jureepon Roboon et al.
Frontiers in cellular neuroscience, 13, 258-258 (2019-06-28)
CD38 is an enzyme that catalyzes the synthesis of cyclic adenosine diphosphate-ribose from nicotinamide adenine dinucleotide (NAD+). We recently reported that this molecule regulates the maturation and differentiation of glial cells such as astrocytes and oligodendrocytes (OLs) in the developing
Ester Orav et al.
Molecular neurobiology, 56(11), 7473-7489 (2019-05-03)
Kainate type ionotropic glutamate receptors (KARs) are expressed in hippocampal interneurons and regulate interneuron excitability and GABAergic transmission. Neuropilin tolloid-like proteins (NETO1 and NETO2) act as KAR auxiliary subunits; however, their significance for various functions of KARs in GABAergic interneurons

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service