Skip to Content
Merck
All Photos(2)

Documents

MAB377A5

Sigma-Aldrich

Anti-NeuN Antibody, clone A60, Alexa Fluor 555 Conjugate

clone A60, from mouse, ALEXA FLUOR 555

Synonym(s):

NEUronal Nuclei, Neuronal Nuclei

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

A60, monoclonal

species reactivity

rat, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

NeuN antibody (NEUronal Nuclei; clone A60) specifically recognizes the DNA-binding, neuron-specific protein NeuN, which is present in most CNS and PNS neuronal cell types of all vertebrates tested. NeuN protein distributions are apparently restricted to neuronal nuclei, perikarya and some proximal neuronal processes in both fetal and adult brain although, some neurons fail to be recognized by NeuN at all ages: INL retinal cells, Cajal-Retzius cells, Purkinje cells, inferior olivary and dentate nucleus neurons, and sympathetic ganglion cells are examples (Mullen et al., 1992; Wolf et al., 1996). Immunohistochemically detectable NeuN protein first appears at developmental timepoints that correspond with the withdrawal of the neuron from the cell cycle and/or with the initiation of terminal differentiation of the neuron (Mullen et al., 1992). Immunoreactivity appears around E9.5 in the mouse neural tube and is extensive throughout the developing nervous system by E12.5. Strong nuclear staining suggests a nuclear regulatory protein function; however, no evidence currently exists as to whether the NeuN protein antigen has a function in the distal cytoplasm or whether it is merely synthesized there before being transported back into the nucleus. No difference between protein isolated from purified nuclei and whole brain extract on immunoblots has been found (Mullen et al., 1992).

Application

Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected NeuN in mouse adult brain cryosections.
This Anti-NeuN Antibody, clone A60, Alexa Fluor 555 Conjugate is validated for use in ICC, IHC for the detection of NeuN.

Quality

Evaluated by Immunocytochemistry in rat E18 primary cortex cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected NeuN in rat E18 primary cortex cells.

Target description

The unconjugated parent antibody (Cat. No. MAB377) has an observed molecular weight in the 46-48 kDa range and possibly another band at approximately 66 kDa

Other Notes

Unconjugated Anti-NeuN has been shown to work with multiple species including Human, Ferret, Chicken, and Avian.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yehe Liu et al.
Biomedical optics express, 12(10), 6571-6589 (2021-11-09)
Axially swept light-sheet microscopy (ASLM) is an effective method of generating a uniform light sheet across a large field of view (FOV). However, current ASLM designs are more complicated than conventional light-sheet systems, limiting their adaptation in less experienced labs.
Ángeles Montilla-García et al.
Neuropharmacology, 125, 231-242 (2017-08-02)
Grip strength deficit is a measure of pain-induced functional disability in rheumatic disease. We tested whether this parameter and tactile allodynia, the standard pain measure in preclinical studies, show parallels in their response to analgesics and basic mechanisms. Mice with
Madeleine Reilly et al.
Scientific reports, 12(1), 9493-9493 (2022-06-11)
There is an urgent need to develop disease-modifying therapies to treat neurodegenerative diseases which pose increasing challenges to global healthcare systems. Prion diseases, although rare, provide a paradigm to study neurodegenerative dementias as similar disease mechanisms involving propagation and spread
Martha E Zeeman et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 34(8), 1439-1446 (2016-08-30)
Whole-body vibration (WBV) is linked epidemiologically to neck and back pain in humans, and to forepaw mechanical allodynia and cervical neuroinflammation in a rodent model of WBV, but the response of the low back and lumbar spine to WBV is
Gregory J Remigio et al.
Neurobiology of disease, 105, 221-234 (2017-06-19)
Memory deficits have a significant impact on the quality of life of patients with epilepsy and currently no effective treatments exist to mitigate this comorbidity. While these cognitive comorbidities can be associated with varying degrees of hippocampal cell death and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service