Skip to Content
Merck
All Photos(1)

Key Documents

MAB3750

Sigma-Aldrich

Anti-LEF-1 Antibody, all isoforms, clone 1C3.1D10

clone 1C3.1D10, Chemicon®, from mouse

Synonym(s):

Lymphoid Enhancer Factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$719.00

$719.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$719.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$719.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C3.1D10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

input

sample type hematopoietic stem cell(s)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LEF1(51176)

General description

Lymphoid Enhancer Factor-1 (LEF-1) is an HMG 1/2-like DNA binding/bending protein and is a member of the LEF/TCF transcription factor family. There are four LEF/TCF family members in mammalian systems (LEF-1, TCF-1, TCF-3 and TCF-4), and orthologs to these factors have been identified in many different species. LEF/TCFs are downstream mediators of Wnt/Wingless signals. Wnt signaling drives cell polarity, cell fate and cell growth decisions in embryonic tissues and in post-natal tissues that continue to develop from mitotically active stem cell precursors. Misregulation of Wnt signaling is also implicated as a root cause of many different cancers, such as colon cancer, melanoma, breast cancer, prostate cancer and others. This antibody recognizes an epitope in amino acids 256-308 of LEF-1. This region is between the transactivation domain and the HMG binding domain of LEF-1 and is conserved in all LEF-1 isoforms.

Specificity

Recognizes human LEF-1 (Lymphoid Enhancer Factor-1) protein. Does not cross-react with TCF-1 or TCF-4.

Immunogen

Epitope: all isoforms
Recombinant full length LEF-1 protein.

Application

Anti-LEF-1 Antibody, all isoforms, clone 1C3.1D10 is an antibody against LEF-1 for use in WB.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells

Hematopoietic Stem Cells
Western blot

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-1130

Physical form

Format: Purified
Purified immunoglobulin. Provided as solution in phosphate buffered saline with 0.08% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Jurkat cell lysate and human colon adenocarcinoma tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service