Skip to Content
Merck
All Photos(1)

Key Documents

MAB366

Sigma-Aldrich

Anti-MAP1B Antibody, clone AA6

ascites fluid, clone AA6, Chemicon®

Synonym(s):

MAP5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$810.00

$810.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$810.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$810.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AA6, monoclonal

species reactivity

feline, chicken, hamster, rat, bovine, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAP1B(4131)

Specificity

Reacts specifically with MAP1B (MAP5) by immunoblot. Shows selective staining of dendritic trees throughout the brain. No cross-reactivity observed to other MAPs or tubulin.

Monoclonal Anti-MAP1b is immunospecific for MAP1b (also known as MAP5, MAPIX or MAP1.2; Riederer et al., 1986; Tucker et al., 1988) as determined by an immunoblot assay. Addition of antibody to microtubule proteins before polymerization results in abnormally short (but otherwise morphologically normal) microtubules.

Immunogen

Rat brain microtubule-associated protein.

Application

Anti-MAP1B Antibody, clone AA6 detects level of MAP1B & has been published & validated for use in WB, IH.
Immunoblotting at 1:300-1:500.

Immunohistochemistry

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Neurofilament & Neuron Metabolism

Physical form

Ascites. Liquid containing 15mM sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Pattnaik et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 20(18), 6789-6796 (2000-09-21)
Protein MAP1B was recently reported to link GABA(C) receptors to the cytoskeleton at neuronal synapses. This interaction was demonstrated in the mammalian retina, where GABA(C) receptors were thought to be exclusively expressed in bipolar cells. Our previous studies on cultured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service